Skip to Content
Merck
All Photos(2)

Documents

C2331

Sigma-Aldrich

Monoclonal Anti-Carcinoembryonic Antigen antibody produced in mouse

clone C6G9, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

C6G9, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:8,000 using human colon carcinoma tissue
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEACAM5(1048)

General description

Carcinoembryonic Antigen (CEA) is a member of the immunoglobulin superfamily that regulates cellular signaling and adhesion. CEA inhibits cell differentiation and subsequently facilitates tumor growth . Thus, CEA has been implicated in various cancers, including colorectal, lung and breast tumors
Monoclonal Anti-Human Carcinoembryonic Antigen (CEA) reacts specifically with human CEA (CD66e, 180 kDa) from different types of malignant tissues. The antibody detects an epitope which is resistant to 30 minute oxidation by 1% sodium periodate solution in formalin-fixed, paraffin-embedded tissue sections. Enzymatic predigestion with proteolytic enzymes enhances immunohistochemical staining with the antibody.The product reacts with medullary thyroid cancers and also associates with cell surface and cytoplasm of malignant glands in colorectal adenocarcinomas. Additionally, the product reacts weakly with normal colon mucosa and occasionally reacts with bile canalicular and pancreatic acinar cells. However, the antibody does not react with other tested tissue such as term placenta, nor does it bind to non-specific cross-reacting antigen (NCA) in granulocytes.
Carcinoembryonic Antigen (CEA) is located on human chromosome 19q13.2.

Specificity

The antibody reacts specifically with human CEA (CD66e, 180 kDa) from several types of malignant tissue including colorectal, lung and breast tumors. It also stains medullary carcinomas of the thyroid. The product reacts strongly with the cell surface and cytoplasm of malignant glands in colorectal adenocarcinomas. A weak reactivity is seen with normal colon mucosa and occasionally with bile canalicular and pancreatic acinar cells, but not with other tested tissue including term placenta. There is no cross-reactivity with non-specific cross-reacting antigen (NCA) present in granulocytes, using immunoblotting, flow cytometry and immunohistochemical techniques. The antibody recognizes an epitope that is resistant to 30 minute oxidation by 1% sodium periodate solution in routine formalin-fixed and paraffin-embedded tissue sections. Enzymatic predigestion with proteolytic enzymes improves immunohistochemical staining with the antibody.

Immunogen

carcinoembryonic antigen (CEA) isolated from a human colon adenocarcinoma cell line.

Application

Monoclonal Anti-Carcinoembryonic Antigen antibody is suitable for use in immunohistochemistry (1:8,000 using formalin-fixed, paraffin-embedded sections of human colon carcinoma tissue).
Monoclonal Anti-Carcinoembryonic Antigen antibody produced in mouse has been used in western blot analysis and surface plasmon resonance (SPR) based immunoassay.

Biochem/physiol Actions

Carcinoembryonic Antigen (CEA) inhibits tumor growth in colon, hepatoma and prostate cancers.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Virna Cortez-Retamozo et al.
Cancer research, 64(8), 2853-2857 (2004-04-17)
Nanobodies are the smallest fragments of naturally occurring single-domain antibodies that have evolved to be fully functional in the absence of a light chain. Nanobodies are strictly monomeric, very stable, and highly soluble entities. We identified a nanobody with subnanomolar
The role of the carcinoembryonic antigen receptor in colorectal cancer progression
Bajenova O, et al.
Journal of integrative oncology., 6(2), 192-192 (2017)
Characterizing the Tumor Suppressor Role of CEACAM1 in Multiple Myeloma
Xu J, et al.
Cellular Physiology and Biochemistry, 45(4), 1631-1640 (2018)
A genetic linkage map of the mouse: current applications and future prospects.
Copeland NG
Science, 262(5130), 57-66 (1993)
Yuuki Shichi et al.
Scientific reports, 9(1), 10871-10871 (2019-07-28)
Pancreatic cancer, composed of heterogeneous cancer cells, alters epithelial to mesenchymal features during growth and metastasis. In this study, we aimed to characterize pancreatic ductal adenocarcinoma (PDAC) cells showing epithelial or mesenchymal features in 3D culture. In 3D culture, PK-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service