Skip to Content
Merck
All Photos(1)

Key Documents

C1743

Sigma-Aldrich

Anti-Cul7 antibody, Mouse monoclonal

clone Ab38, purified from hybridoma cell culture

Synonym(s):

Anti-Cullin 7, Anti-KIAA007

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ab38, monoclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa

species reactivity

human, mouse

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 0.5-1 μg/mL using 3T3 total cel extract

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUL7(9820)
mouse ... Cul7(66515)

General description

Monoclonal Anti-Cul7 (mouse IgG2b isotype) is derived from the hybridoma Ab38 produced by the fusion of mouse myeloma cells (P3X63Ag8.653) and splenocytes from BALB/c mice immunized with mouse Cul7. CUL7 is expressed in all tissues and cell lines except testis and small intestine.

Immunogen

mouse Cul7.

Application

Monoclonal Anti-Cul7 antibody produced in mouse is suitable for immunoprecipitation and western blotting at a concentration of 0.5-1μg/mL using 3T3 total cel extract.

Biochem/physiol Actions

CUL7 functions to promote cell growth through and antagonizes the function of p53.
Cullin-7 (cul7) is a protein encoded by the CUL7 gene in humans. It is also referred as 3M1, KIAA0076 and dJ20C7.5. Mutation in CUL7 gene is majorly responsible for 3M syndrome (is a rare autosomal recessive disorder). CUL7 acts as a novel gene potentially involved in liver carcinogenesis associated with metabolic syndrome (MS) and its amplification may influence cell proliferation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytoplasmic localized ubiquitin ligase cullin 7 binds to p53 and promotes cell growth by antagonizing p53 function
Andrews P, et al.
Oncogene, 25(33), 4534-4534 (2006)
Melanie Anger et al.
PloS one, 15(12), e0244096-e0244096 (2020-12-23)
Fibrosis is a pathognomonic feature of structural heart disease and counteracted by distinct cardioprotective mechanisms, e.g. activation of the phosphoinositide 3-kinase (PI3K) / AKT pro-survival pathway. The Cullin-RING E3 ubiquitin ligase 7 (CRL7) was identified as negative regulator of PI3K/AKT
Céline Huber et al.
Best practice & research. Clinical endocrinology & metabolism, 25(1), 143-151 (2011-03-15)
3M syndrome (MIM 273750) is an autosomal recessive disorder characterized by pre- and post-natal growth retardation (<-4 SD), facial dysmorphism, large head circumference, normal intelligence and endocrine function. Skeletal changes include long slender tubular bones and tall vertebral bodies. There
Céline Huber et al.
European journal of human genetics : EJHG, 17(3), 395-400 (2009-02-20)
The 3M syndrome is a rare autosomal recessive disorder recently ascribed to mutations in the CUL7 gene and characterized by severe pre- and postnatal growth retardation. Studying a series of 33 novel cases of 3M syndrome, we have identified deleterious
Valerie Paradis et al.
Gut, 62(6), 911-919 (2012-09-04)
Metabolic syndrome (MS) is an emerging risk factor in hepatocellular carcinoma (HCC). HCC related to MS may occur either in advanced fibrosis or before the development of cirrhosis, suggesting involvement of different molecular pathways according to the features of background

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service