Skip to Content
Merck

Skip To

C1506

Cytidine 5′-triphosphate disodium salt

≥95%, P2X4 purinergic receptor agonist, powder

Synonym(s):

CTP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H14N3Na2O14P3
CAS Number:
Molecular Weight:
527.12
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
252-849-3
MDL number:
Assay:
≥95%
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cytidine 5′-triphosphate disodium salt, ≥95%

Quality Segment

assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O

InChI

1S/C9H16N3O14P3.2Na/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H2,10,11,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1

InChI key

NFQMDTRPCFJJND-WFIJOQBCSA-L

General description

Cytidine 5′-triphosphate is a nucleoside triphosphate with a pyrimidine ring. It is present in RNA.

Application

Cytidine 5′-triphosphate disodium salt has been used:
  • for the conversion of adenosine monophosphate (AMP) to adenosine diphosphate (ADP) in luminometry analysis
  • in the preparation of ribonucleotide 5′-triphosphate precursors stocks for large-scale RNA synthesis
  • to perform in vitro transcription

Biochem/physiol Actions

Cytidine 5′-triphosphate (CTP) prevents the action of aspartate carbamoyltransferase. This enzyme participates in pyrimidine biosynthesis. Like adenosine triphosphate (ATP), CTP serves as a molecule of high energy. It acts as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation.
P2X4 purinergic receptor agonist.

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C855230320C9755
description

≥95%

description

HPLC purified, aqueous solution for RNA polymerase transcription

description

≥90.0% (HPLC)

description

from yeast, ≥95%

assay

≥95%

assay

-

assay

≥90.0% (HPLC)

assay

≥95%

form

powder

form

liquid

form

solid

form

powder

Quality Level

200

Quality Level

300

Quality Level

100

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters