Skip to Content
Merck
All Photos(3)

Documents

C104

Sigma-Aldrich

(±)-CPP

solid

Synonym(s):

(±)-3-(2-Carboxypiperazin-4-yl)propyl-1-phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N2O5P
CAS Number:
Molecular Weight:
252.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

H2O: 99.2 mg/mL
ethanol: insoluble

SMILES string

OC(=O)C1CN(CCCP(O)(O)=O)CCN1

InChI

1S/C8H17N2O5P/c11-8(12)7-6-10(4-2-9-7)3-1-5-16(13,14)15/h7,9H,1-6H2,(H,11,12)(H2,13,14,15)

InChI key

CUVGUPIVTLGRGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(±)-CPP has been used as an uncompetitive NMDA receptor antagonist:
  • to study its effects on blood glucose in mice
  • in electrophysiological recording in mouse brain slices
  • in electrophysiological recording in rat hippocampal pyramidal neurons

Biochem/physiol Actions

3-(2-carboxypiperazin-4yl) propyl-1-phosphonic acid (CPP) is a potent, competitive, and selective N-methyl-D-aspartate (NMDA) receptor antagonist. It is an analog of 2-amino-7-phosphonoheptanoic acid. CPP shows anticonvulusant and antinociceptive activity.

Caution

Hygroscopic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammadali Foroozandeh et al.
Magnetic resonance in chemistry : MRC, 53(11), 894-900 (2015-08-21)
Resolution enhancement is a long-sought goal in NMR spectroscopy. In conventional multidimensional NMR experiments, such as the (1) H-(13) C HSQC, the resolution in the indirect dimensions is typically 100 times lower as in 1D spectra because it is limited
Thien-Tri Cung et al.
The New England journal of medicine, 373(11), 1021-1031 (2015-09-01)
Experimental and clinical evidence suggests that cyclosporine may attenuate reperfusion injury and reduce myocardial infarct size. We aimed to test whether cyclosporine would improve clinical outcomes and prevent adverse left ventricular remodeling. In a multicenter, double-blind, randomized trial, we assigned
Neil Schwartz et al.
Neuron, 70(3), 455-467 (2011-05-11)
In the developing Xenopus tadpole, conditioning with 20 min of visual stimulation leads to increased proBDNF protein levels in the tectum measured 4 hr later. Following conditioning, the ability to induce direction selectivity in tectal neurons, as well as both
Wayne W Tschetter et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(12), 5362-5366 (2013-03-22)
We previously reported in adult mice that visuomotor experience during monocular deprivation (MD) augmented enhancement of visual-cortex-dependent behavior through the non-deprived eye (NDE) during deprivation, and enabled enhanced function to persist after MD. We investigated the physiological substrates of this
Marcel B M Teunissen et al.
The Journal of investigative dermatology, 134(12), 2898-2907 (2014-06-20)
IL-17A is pivotal in the etiology of psoriasis, and CD8(+) T cells with the ability to produce this cytokine (Tc17 cells) are over-represented in psoriatic lesions. Here we demonstrate that the frequency of Tc17 cells in peripheral blood of psoriasis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service