Skip to Content
Merck
All Photos(3)

Key Documents

C0871

Sigma-Aldrich

Anti-Cullin 3 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CUL3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

rat, mouse, human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysate of rat brain
indirect immunofluorescence: 5-10 μg/mL using mouse 3T3 cells
western blot: 1-2 μg/mL using whole extract of HEK-293T cells expressing human Cullin 3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUL3(8452)
mouse ... Cul3(26554)
rat ... Cul3(301555)

Related Categories

General description

Cullin 3 is a member of the cullin family of E3 ubiquitin protein ligases. Cullin 3 forms the BTB-Cul3-Rbx1 (BCR) ubiquitin ligase complex that contains a RING-box protein, Rbx1, and a BTB containing protein. Rbx1 binds to the C-terminus of cullin 3 through its RING-type zinc finger domain to recruit a ubiquitin-conjugating enzyme. The BTB containing protein binds cullin 3 at its N-terminus and functions as a cullin specific adaptor protein.

Application

Anti-Cullin 3 antibody produced in rabbit has been used in
  • western blotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Cullin 3 (Cul3) acts as a key mediator of post-translational protein regulation. It functions as a scaffold protein that assemble multi-subunit ubiquitin ligase complexes which are responsible for the specific recognition and targeting of substrates for ubiquitin-dependent degradation by the 26S proteasome. Cul3 is required for degradation of the microtubule-severing protein MEI-1/katanin during transitioning from meiosis-to-mitosis phase. It forms a catalytically inactive BTB-Cul3-Rbx1 (BCR) ubiquitin ligase and becomes functional upon covalent attachment of the ubiquitin homologue neural-precursor-cell-expressed. BCR ubiquitin ligase down regulated 8 (Nedd8) near the C terminus of Cul3.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Missense variants in RHOBTB2 cause a developmental and epileptic encephalopathy in humans, and altered levels cause neurological defects in Drosophila
Straub J, et al.
American Journal of Human Genetics, 102(1), 44-57 (2018)
Lionel Pintard et al.
The EMBO journal, 23(8), 1681-1687 (2004-04-09)
Cullin-based E3 ligases target substrates for ubiquitin-dependent degradation by the 26S proteasome. The SCF (Skp1-Cul1-F-box) and ECS (ElonginC-Cul2-SOCS box) complexes are so far the best-characterized cullin-based ligases. Their atomic structure has been solved recently, and several substrates have been described
Matthew D Petroski et al.
Nature reviews. Molecular cell biology, 6(1), 9-20 (2005-02-03)
Cullin-RING complexes comprise the largest known class of ubiquitin ligases. Owing to the great diversity of their substrate-receptor subunits, it is possible that there are hundreds of distinct cullin-RING ubiquitin ligases in eukaryotic cells, which establishes these enzymes as key
Wananit Wimuttisuk et al.
Molecular biology of the cell, 18(3), 899-909 (2006-12-29)
Cullins are members of a family of scaffold proteins that assemble multisubunit ubiquitin ligase complexes to confer substrate specificity for the ubiquitination pathway. Cullin3 (Cul3) forms a catalytically inactive BTB-Cul3-Rbx1 (BCR) ubiquitin ligase, which becomes functional upon covalent attachment of
Lionel Pintard et al.
Nature, 425(6955), 311-316 (2003-09-19)
Many biological processes, such as development and cell cycle progression are tightly controlled by selective ubiquitin-dependent degradation of key substrates. In this pathway, the E3-ligase recognizes the substrate and targets it for degradation by the 26S proteasome. The SCF (Skp1-Cul1-F-box)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service