Skip to Content
Merck

Skip To

C0412

Concanavalin A from Canavalia ensiformis (Jack bean)

Type IV-S, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture

Synonym(s):

Con A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
23151817
EC Number:
234-258-2
NACRES:
NA.32
MDL number:
Form:
lyophilized powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

γ-irradiated

type

Type IV-S

product line

BioReagent

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

impurities

salt, essentially free

solubility

PBS: 5 mg/mL, slightly hazy, H2O: soluble, slightly hazy

storage temp.

−20°C

Application

Concanavalin A from Canavalia ensiformis (jack bean) has been used:
  • to induce spleen blastogenesis
  • to generate ConA blast targets from B6 and lpr spleen cells
  • to stimulate splenic lymphocytes from estrogen-and placebo-treated mice

Biochem/physiol Actions

Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Ca2+ and Mn2+ ions are required for activity. Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer; above pH 7.0 higher aggregates are formed. Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6.

Analysis Note

Where reported, agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, containing Ca2+ and Mn2+ of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C5275L7647L6397
description

Type IV-S, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture

description

Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture

description

Type VI, lyophilized powder

description

peroxidase conjugate, lyophilized powder

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

blood typing: suitable, fractionation: suitable

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

H2O: soluble, slightly hazy, PBS: 5 mg/mL, slightly hazy

solubility

H2O: soluble, slightly hazy, PBS: 5 mg/mL, slightly hazy

solubility

water: 1 % (w/v)

solubility

-

impurities

salt, essentially free

impurities

salt (Contains buffer salts and NaCl)

impurities

Carbohydrate, essentially free, salt, essentially free

impurities

carbohydrate, essentially free

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. ConA 제품 (C0412)에 관해 질문 드립니다. ConA가 Ca2+, Mn2+ 이온이 존재해야 활성화가 된다고 하는데 해당 제품은 powder형태이며 이는 따로 용액이 필요합니다. 그렇다면 이 제품 안에 저 금속 이온이 함께 들어있는 건지, 아니면 Sigma 제품 중에 이 제품을 혼합할 수 있는 PBS가 따로 있는지(있다면 카탈로그 번호 알려주셨으면 합니다) 궁금합니다.

    1 answer
    1. Concanavalin A has 2 metal binding sites per monomer. Calcium and Manganese are required to bind these sites in order for sugar binding to occur. It is recommended to prepare stock solutions in PBS containing Ca2+ and Mn2+.

      See the link below to the product datasheet which includes preparation instructions:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/331/341/c0412dat.pdf

      Helpful?

Reviews

No rating value

Active Filters