Skip to Content
Merck
All Photos(1)

Key Documents

C0167

Sigma-Aldrich

Calcitonin Gene Related Peptide human

≥95% (HPLC), powder

Synonym(s):

α-CGRP, CGRP-I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C163H267N51O49S2
CAS Number:
Molecular Weight:
3789.31
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

SC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)C)CS)CC(=O)O)[C@H](O)C)C)[C@H](O)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H]([C@H](O)C)C(=O)N[C@@H](Cc4nc[nH]c4)C(=O)N[C@@H](CCCNC(=N)N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N

InChI

1S/C163H269N51O49S2/c1-73(2)52-97(186-116(226)65-179-131(233)82(18)183-139(241)98(53-74(3)4)193-137(239)94(44-35-49-176-162(171)172)188-142(244)101(57-91-62-175-72-182-91)199-159(261)128(88(24)221)213-156(258)123(79(13)14)207-151(253)110(71-265)204-160(262)126(86(22)219)210-133(235)84(20)185-157(259)125(85(21)218)211-147(249)105(61-119(229)230)198-150(252)109(70-264)203-130(232)81(17)166)140(242)194-99(54-75(5)6)141(243)202-108(69-217)149(251)190-95(45-36-50-177-163(173)174)138(240)201-106(67-215)134(236)180-63-115(225)178-64-118(228)205-121(77(9)10)155(257)208-122(78(11)12)154(256)191-93(43-32-34-48-165)136(238)196-102(58-112(167)222)144(246)197-103(59-113(168)223)143(245)195-100(56-90-40-29-26-30-41-90)145(247)209-124(80(15)16)161(263)214-51-37-46-111(214)152(254)212-127(87(23)220)158(260)200-104(60-114(169)224)146(248)206-120(76(7)8)153(255)181-66-117(227)187-107(68-216)148(250)189-92(42-31-33-47-164)135(237)184-83(19)132(234)192-96(129(170)231)55-89-38-27-25-28-39-89/h25-30,38-41,62,72-88,92-111,120-128,215-221,264-265H,31-37,42-61,63-71,164-166H2,1-24H3,(H2,167,222)(H2,168,223)(H2,169,224)(H2,170,231)(H,175,182)(H,178,225)(H,179,233)(H,180,236)(H,181,255)(H,183,241)(H,184,237)(H,185,259)(H,186,226)(H,187,227)(H,188,244)(H,189,250)(H,190,251)(H,191,256)(H,192,234)(H,193,239)(H,194,242)(H,195,245)(H,196,238)(H,197,246)(H,198,252)(H,199,261)(H,200,260)(H,201,240)(H,202,243)(H,203,232)(H,204,262)(H,205,228)(H,206,248)(H,207,253)(H,208,257)(H,209,247)(H,210,235)(H,211,249)(H,212,254)(H,213,258)(H,229,230)(H4,171,172,176)(H4,173,174,177)/t81-,82-,83-,84-,85+,86+,87+,88+,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,120-,121-,122-,123-,124-,125-,126-,127-,128-/m0/s1

InChI key

DNKYDHSONDSTNJ-XJVRLEFXSA-N

Gene Information

human ... CALCA(796)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Cys-Asp-Thr-Ala-Thr-Cys-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asn-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2 [Disulfide Bridge: 2-7]

Biochem/physiol Actions

Potent, long-lasting vasodilator.
Potent, long-lasting vasodilator; activation of CGRP receptors on pancreatic β-cells increases plasma levels of pancreatic enzymes.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivia Benguettat et al.
PLoS pathogens, 14(9), e1007279-e1007279 (2018-09-05)
The digestive tract is the first organ affected by the ingestion of foodborne bacteria. While commensal bacteria become resident, opportunistic or virulent bacteria are eliminated from the gut by the local innate immune system. Here we characterize a new mechanism
Somayeh Shahrokhi et al.
Growth factors (Chur, Switzerland), 28(1), 66-73 (2009-11-17)
There is little evidence on roles of growth factors other than cytokines in expansion of cord blood (CB) stem cells. We aimed to explore a novel approach for expansion, using Substance P (SP) and calcitonin gene-related peptide (CGRP) neuropeptides. CB
Lu Yu et al.
Molecular pain, 4, 61-61 (2008-12-06)
The present study aims to investigate the role of transient receptor potential vanilloid 1 (TRPV1) in dorsal root ganglion (DRG) neurons in chronic pain including thermal hyperalgesia and mechanical allodynia. Chronic inflammatory nociception of rats was produced by intraplantar injection
Amelia Franke-Radowiecka et al.
Journal of anatomy, 235(5), 1007-1017 (2019-07-28)
This study investigated the innervation of internal genital organs in 5-, 7- and 10-week-old female pig foetuses using single and double-labelling immunofluorescence methods. The structure and topography of the organs was examined using a scanning electron microscope (SEM). The investigations
Hui Wang et al.
Arthritis research & therapy, 17, 4-4 (2015-01-13)
Developmental dysplasia of the hip (DDH) is a common musculoskeletal disorder that has pain and loss of joint function as major pathological features. In the present study, we explored the mechanisms of possible involvement and regulation of substance P (SP)

Questions

  1. Hello, I wanted to know what the appropriate diluent is for reconstituting CGRP Cat#C0167. Thanks, Gargi

    1 answer
    1. As listed in the specification sheet for Product No. C0167, Calcitonin Gene Related Peptide human, ≥95% (HPLC) powder is soluble in water or 1% acetic acid (aq.) within a range of 1.00-1.04 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service