BAF378600001
Deepwell 96 well plate
polypropylene, sterile, pack of 24 ea
Synonym(s):
deepwell multiplate, storage plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122102
NACRES:
NB.22
material:
conical bottom clear polypropylene wells (translucent)
polypropylene
polypropylene
size:
96 wells
Recommended Products
material
conical bottom clear polypropylene wells (translucent)
polypropylene
sterility
sterile
packaging
pack of 24 ea
manufacturer/tradename
Bel-Art F37860-0001
size
96 wells
well capacity
2 mL
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These high-form polypropylene 96 well plates expand the flexibility of multichannel or automated pipetting systems. Use for non-binding protein assays, enzyme or receptor assays, or with cell harvesters. The 2 mL per well capacity makes it possible to do serial dilution procedures that are impossible with normal multiwell plates.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ruth J Napier et al.
Nature communications, 11(1), 5406-5406 (2020-10-28)
Mutations in nucleotide-binding oligomerization domain-containing protein 2 (NOD2) cause Blau syndrome, an inflammatory disorder characterized by uveitis. The antimicrobial functions of Nod2 are well-established, yet the cellular mechanisms by which dysregulated Nod2 causes uveitis remain unknown. Here, we report a
Nils T Vethe et al.
Therapeutic drug monitoring, 41(5), 607-614 (2019-10-05)
Therapeutic drug monitoring is standard practice for the immunosuppressant tacrolimus (Tac). Venous blood sampling at outpatient clinics is time-consuming and impractical with regard to obtaining trough concentrations on clinical visit days. Home-based blood sampling may be patient friendly and pave
Nataša Mehle et al.
Plants (Basel, Switzerland), 9(3) (2020-03-08)
In recent years, pepino mosaic virus (PepMV) has rapidly evolved from an emerging virus to an endemic pathogen, as it causes significant loses to tomato crops worldwide. At present, the main control strategy for prevention of PepMV disease in tomato
Philip Storck et al.
Antibiotics (Basel, Switzerland), 9(12) (2020-12-02)
The increasing number of infections caused by multidrug-resistant bacteria requires an intensified search for new antibiotics. Pep19-4LF is a synthetic antimicrobial peptide (GKKYRRFRWKFKGKLFLFG) that was previously designed with the main focus on high antimicrobial activity. The hydrophobic motif, LFLFG, was
Alison E John et al.
Nature communications, 11(1), 4659-4659 (2020-09-18)
The αvβ6 integrin plays a key role in the activation of transforming growth factor-β (TGFβ), a pro-fibrotic mediator that is pivotal to the development of idiopathic pulmonary fibrosis (IPF). We identified a selective small molecule αvβ6 RGD-mimetic, GSK3008348, and profiled
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service