Skip to Content
Merck

B9936

Boc-Ile-Glu-Gly-Arg-7-amido-4-methylcoumarin acetate salt

≥97% (HPLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H50N8O10
CAS Number:
Molecular Weight:
730.81
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Boc-Ile-Glu-Gly-Arg-7-amido-4-methylcoumarin acetate salt, ≥97% (HPLC)

assay

≥97% (HPLC)

form

powder

solubility

methanol: 2 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

N(C(CCCN=C(N)N)C(=O)NC(=O)CNC(=O)C(NC(=O)C(NC(=O)OC(C)(C)C)C(CC)C)CCC(=O)O)c1cc2c(cc1)C(=CC(=O)O2)C

InChI

1S/C34H50N8O10/c1-7-18(2)28(42-33(50)52-34(4,5)6)31(49)40-23(12-13-26(44)45)29(47)38-17-25(43)41-30(48)22(9-8-14-37-32(35)36)39-20-10-11-21-19(3)15-27(46)51-24(21)16-20/h10-11,15-16,18,22-23,28,39H,7-9,12-14,17H2,1-6H3,(H,38,47)(H,40,49)(H,42,50)(H,44,45)(H4,35,36,37)(H,41,43,48)

InChI key

PWOIMWXLMWHALO-UHFFFAOYSA-N

General description

A sensitive and specific fluorogenic substrate for factor Xa.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library