Skip to Content
Merck

B9685

BAY 61-3606 hydrochloride hydrate

≥98% (HPLC), powder

Synonym(s):

2-[[7-(3,4-Dimethoxyphenyl)imidazo[1,2-c]pyrimidin-5-yl]amino]pyridine-3-carboxamide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H18N6O3 · xHCl · yH2O
Molecular Weight:
390.40 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

potency

7.5 nM Ki

storage condition

desiccated

color

yellow

solubility

H2O: soluble

originator

Bayer

storage temp.

2-8°C

SMILES string

O.Cl.COc1ccc(cc1OC)-c2cc3nccn3c(Nc4ncccc4C(N)=O)n2

InChI

1S/C20H18N6O3.ClH.H2O/c1-28-15-6-5-12(10-16(15)29-2)14-11-17-22-8-9-26(17)20(24-14)25-19-13(18(21)27)4-3-7-23-19;;/h3-11H,1-2H3,(H2,21,27)(H,23,24,25);1H;1H2

InChI key

WYXPKEXHEJAVSI-UHFFFAOYSA-N

Application

BAY 61-3606 hydrochloride hydrate has been used in focal cerebral ischemia model. It has also been used to assess the effect of Syk in stroke-induced injury.

Biochem/physiol Actions

Spleen tyrosine kinase (Syk) inhibitor; anti-inflammatory. Syk plays a major role in inflammation pathways via receptors for the Fc portion of immunoglobulins, as well as B cell signaling. It is orally active, potent (Ki 7.5 nM), and highly selective for Syk vs other kinases (Lyn, Fyn, Src, Itk, Btk; Ki′s).
BAY 61-3606 helps to prevent the degranulation of freshly isolated basophils. It also prevents B cell receptor (BCR)-mediated signaling.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library