Skip to Content
Merck

B9183

Anti-HA-Biotin antibody, Mouse monoclonal

clone HA-7, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-HA, Monoclonal Anti-HA-Biotin antibody produced in mouse, Anti-HA, Anti-Influenza Hemagglutinin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.56
MDL number:
Conjugate:
biotin conjugate
Clone:
HA-7, monoclonal
Application:
WB CL
Citations:
7
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

biotin conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HA-7, monoclonal

form

buffered aqueous solution

concentration

~1 mg/mL

technique(s)

western blot (chemiluminescent): 0.25-0.50 μg/mL using HA tagged fusion proteins in transiently transfected mammalian cell extracts.

shipped in

dry ice

storage temp.

2-8°C

General description

The Monoclonal Anti-HA Biotin Conjugate antibody recognizes the HA tag sequence on the HA tagged fusion proteins when expressed at the N or C terminal to the fusion protein. This antibody is derived from the HA-7 hybridoma, which is produced from the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to the amino acid residues 98-106 (YPYDVPDYA) of human Influenza virus hemagglutinin (HA) known as HA-tag, conjugated to KLH. Ascites fluid collected from the HA-7 hybridoma is then conjugated to biotin.

Immunogen

synthetic peptide corresponding to a fragment of human influenza virus hemagglutinin (HA) known as HA-tag, conjugated to KLH

Application

Antibody suitable for immunoblotting.

Biochem/physiol Actions

The antibody recognizes native as well as denatured-reduced forms of HA-tagged proteins and is reactive with N- or C-terminal HA-tagged fusion proteins expressed in E. coli or in mammalian cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library