B9157
Nalgene® bottles, style 2104
capacity 250 mL
Synonym(s):
HDPE bottles, nalgene bottle, nalgene bottles
About This Item
Recommended Products
material
polypropylene cap
polypropylene screw closure
round bottle
translucent high-density polyethylene bottle
Joint
Joints threaded neck (43)
Agency
suitable for EPA OTM-45
description
Not autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2104-0008
bottle diam. × H
61 mm × 131 mm
capacity
250 mL
application(s)
PFAS testing
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Nalgene® bottles are ideal for storing chemicals, materials, and specimens.
- Laboratory-grade HDPE material for reliably low leachables and extractables.
- Suitable for use in freezers (up to -100°C).
- Rigid, uniform walls offer protection against splitting or punctures.
- Bottles have excellent durability and are chemically resistant to most acids, bases, and alcohols.
- Linerless caps ensure a leak-proof seal for sample protection.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service