Skip to Content

B8276

Scienceware® biohazard disposal bags

W × H × thickness 25 in. (64 cm) × 35 in. (89 cm) × 0.002 in., red, pack of 200

Synonym(s):

autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags, 131642535, F13164-2535

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
24111500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Imprinted in black with biohazard symbol and precautions in English, Spanish, French, and German.

packaging

pack of 200

manufacturer/tradename

Bel-Art F13164-2535

W × H × thickness

25 in. (64 cm) × 35 in. (89 cm) × 0.002 in.

color

red

Features and Benefits

Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • High temperature steam autoclavable to 135˚C (275˚F)

Legal Information

Scienceware is a registered trademark of Bel-Art Products, a division of SP


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library