Skip to Content

B8263

Benzphetamine hydrochloride

Synonym(s):

N-Benzyl-N,α-dimethylphenethylamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H21N · HCl
CAS Number:
Molecular Weight:
275.82
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352116
EC Number:
226-489-2
MDL number:
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Segment

drug control

USDEA Schedule III; Home Office Schedule 3; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

SMILES string

Cl.C[C@@H](Cc1ccccc1)N(C)Cc2ccccc2

InChI

1S/C17H21N.ClH/c1-15(13-16-9-5-3-6-10-16)18(2)14-17-11-7-4-8-12-17;/h3-12,15H,13-14H2,1-2H3;1H/t15-;/m0./s1

InChI key

ANFSNXAXVLRZCG-RSAXXLAASA-N

Gene Information

human ... SLC18A2(6571)

Biochem/physiol Actions

Adrenergic receptor agonist; CNS stimulant.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


Still not finding the right product?

Explore all of our products under Benzphetamine hydrochloride


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library