Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Conjugate:
biotin conjugate
Clone:
polyclonal
Application:
ELISA (d), IHC (p), WB CL
Citations:
24
biological source
goat
Quality Level
conjugate
biotin conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
direct ELISA: 1:160,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400, western blot (chemiluminescent): 1:300,000 using total cell extract of HeLa cells (5-10 μg/well)
storage temp.
−20°C
target post-translational modification
unmodified
General description
Goat polyclonal anti-Mouse IgG (Fc specific)–Biotin reacts with mouse IgG and mouse IgG, Fc fragment. It shows no reactivity with mouse IgG Fab fragment, human IgG, IgA, IgM, kappa and lambda light chain, bovine IgG and IgM, or horse IgG.
Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.
Biotin:avidin second antibody signal systems involve the conjugation of biotin to a secondary antibody. Once the secondary antibody binds to a target primary antibody in an immunochemical assay, the signal is developed by allowing avidin which has been conjugated to an enzyme or signal molecule to bind to the antibody associated biotin molecule.
Biotin:avidin second antibody signal systems involve the conjugation of biotin to a secondary antibody. Once the secondary antibody binds to a target primary antibody in an immunochemical assay, the signal is developed by allowing avidin which has been conjugated to an enzyme or signal molecule to bind to the antibody associated biotin molecule.
Application
Anti-Mouse IgG (Fc specific)−Biotin antibody produced in goat has been used in ELISA and quantitative suspension array technology (qSAT) multiplex antibody assay.
Goat polyclonal anti-Mouse IgG (Fc specific)–Biotin may be used to detect mouse IgG within serum or other biological systems. The antibody may be used as a secondary antibody in immunochemical assays based on biotin:avidin second antibody signal systems wherein mouse IgG is the primary antibody.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Preparation Note
Adsorbed to reduce background staining with bovine, horse or human samples.
Other Notes
Antibody adsorbed with bovine, equine and human serum proteins.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat
— or —
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The context of tetanus toxoid application influences the outcome of antigen-specific and self-directed humoral immune response
Stojanovic M, et al.
Microbiology and Immunology, 53(2), 89-100 (2009)
Masahiro Kozuka et al.
Cancers, 13(19) (2021-10-14)
Circulating tumor cells (CTCs) are a prognostic marker in patients with metastatic colorectal cancer (mCRC). However, little is known about the characterization of CTCs in mCRC at the single-cell level using RNA sequencing. The purpose of this study was to
Adjuvant dependence of APS pathology-related low-affinity antibodies during secondary immune response to tetanus toxoid in BALB/c mice
Zivkovic
Immunologic Research, 56(1), 143-149 (2013)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| B7401-.5ML | 04061838352682 |
| B7401-1ML | 04061837514999 |