Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
-
Species reactivity:
-
Application:
—
Citations:
1
form
solution
Quality Segment
concentration
5 mg/mL in SDS-PAGE loading buffer
suitability
suitable for positive control in immunoblotting techniques
storage temp.
−20°C
Application
Control for Immunoblotting (recommended use: 5 μl (25 μg) per gel lane)
Other Notes
Whole tissue extract of rat brain.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1
Storage Class
10 - Combustible liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


