Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Pricing and availability is not currently available.
Product Name
Bisdemethoxycurcumin, ≥98% (HPLC), solid
Quality Segment
assay
≥98% (HPLC)
form
solid
solubility
DMSO: ≥20 mg/mL
storage temp.
2-8°C
SMILES string
Oc1ccc(cc1)\C=C\C(=O)CC(=O)\C=C\c2ccc(O)cc2
InChI
1S/C19H16O4/c20-16-7-1-14(2-8-16)5-11-18(22)13-19(23)12-6-15-3-9-17(21)10-4-15/h1-12,20-21H,13H2/b11-5+,12-6+
InChI key
PREBVFJICNPEKM-YDWXAUTNSA-N
Application
Bisdemethoxycurcumin has been used:
- to test its inhibitory effect on cell cycle and mitochondrial function in gastric adenocarcinoma cells
- to test it neuroprotective role against lead (Pb) induced toxicity in dopaminergic and noradrenergic systems of Meriones shawi
- as a standard for calibration curve generation to quantify plasma curcuminoids using high-performance liquid chromatography-diode array detection (HPLC-DAD) and ultraviolet (UV)
Biochem/physiol Actions
Bisdemethoxycurcumin (BMDC) is a stable dimethoxy derivative of curcumin and is useful as a supplement in cell culture medium. It also possesses antimicrobial, antioxidative and neuroprotective functionality.
Bisdemethoxycurcumin (BDMC) is a derivative or curcumin, and represents one of the major active components of curcumin products isolated from Curcumae sp. BDMC shares similar anti-inflammatory properties with demethoxycurcumin. It inhibits LPS-induced nitric oxide (NO) production and expression of iNOS and COX2 in RAW264.7 cells by blocking NF-kB activation. BDMC also displays unique properties in that it enhances Abeta clearance by upregulating expression MGAT3 and TLR genes. BDMC potently inhibits AKR1B10.
Inhibitor of NF-kB transcriptional activity
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC), solid | description ≥98% (HPLC) | description analytical standard | description ≥98% (HPLC), solid |
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥95.0% (HPLC) | assay ≥98% (HPLC) |
| form solid | form powder | form - | form solid |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C |
| solubility DMSO: ≥20 mg/mL | solubility DMSO: 1 mg/mL, clear (warmed) | solubility - | solubility DMSO: >20 mg/mL |
Still not finding the right product?
Explore all of our products under Bisdemethoxycurcumin
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


