Skip to Content
Merck

B6008

Ponceau BS

Dye content ~60 %, Powder

Synonym(s):

Acid Red 66, Biebrich scarlet WS, Biebrich scarlet sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H14N4Na2O7S2
CAS Number:
Molecular Weight:
556.48
UNSPSC Code:
12171500
NACRES:
NA.47
PubChem Substance ID:
EC Number:
224-084-5
MDL number:
Colour Index Number:
26905
Beilstein/REAXYS Number:
3883900
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ponceau BS, Dye content ≥55 %

form

powder

Quality Segment

composition

Dye content, ≥55%

technique(s)

microbe id | staining: suitable

color

red to brown

solubility

ethanol: 1 mg/mL, H2O: >30 mg/mL

εmax

≥19000 at 510 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1ccc2ccccc2c1\N=N\c3ccc(cc3S([O-])(=O)=O)\N=N\c4ccc(cc4)S([O-])(=O)=O

InChI

1S/C22H16N4O7S2.2Na/c27-20-12-5-14-3-1-2-4-18(14)22(20)26-25-19-11-8-16(13-21(19)35(31,32)33)24-23-15-6-9-17(10-7-15)34(28,29)30;;/h1-13,27H,(H,28,29,30)(H,31,32,33);;/q;2*+1/p-2/b24-23+,26-25+;;

InChI key

VVAVKBBTPWYADW-RVTJCSDESA-L

Application

Ponceau BS has been used in Masson′s trichrome stain for the staining of collagen. It has also been used for eosinophil staining.

Biochem/physiol Actions

Ponceau BS is used in Masson′s trichrome staining procedure, along with acid fuchsin, for the labeling of collagen and muscle red.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library