Skip to Content
Merck

Skip To

B5768

Biotin 4-amidobenzoic acid sodium salt

≥95%

Synonym(s):

N-(+)-Biotinyl-4-aminobenzoic acid sodium salt, N-Biotinyl-p-aminobenzoic acid sodium salt, Sodium N-(+)-biotinyl-4-aminobenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H20N3NaO4S
CAS Number:
Molecular Weight:
385.41
NACRES:
NA.46
PubChem Substance ID:
UNSPSC Code:
12352203
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

[Na+].[H][C@]12CS[C@@H](CCCCC(=O)Nc3ccc(cc3)C([O-])=O)[C@@]1([H])NC(=O)N2

InChI

1S/C17H21N3O4S.Na/c21-14(18-11-7-5-10(6-8-11)16(22)23)4-2-1-3-13-15-12(9-25-13)19-17(24)20-15;/h5-8,12-13,15H,1-4,9H2,(H,18,21)(H,22,23)(H2,19,20,24);/q;+1/p-1/t12-,13-,15-;/m0./s1

InChI key

AMXZKFYBAOWERX-HZPCBCDKSA-M

General description

Biotin 4-amidobenzoic acid is a substrate of biotinidase.

Application

Biotin 4-amidobenzoic acid sodium salt has been used as a biotin amide substrate to measure biotinyl-6-aminoquinoline (BAQ) hydrolysis by competitive inhibition.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14407A6928870285P
description

≥95%

description

≥97.0% (HPLC)

description

≥99%, powder

description

Avanti Research - A Croda Brand 870285P, powder

assay

≥95%

assay

≥97.0% (HPLC)

assay

≥99%

assay

>99% (TLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

−20°C

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters