Skip to Content
Merck
All Photos(1)

Documents

B5282

Sigma-Aldrich

Bold Modified Basal Freshwater Nutrient Solution

50 ×, liquid, suitable for plant cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
10171502
NACRES:
NA.72

sterility

sterile-filtered

Quality Level

form

liquid

concentration

50 ×

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

Application

Bold Modified Basal Freshwater Nutrient Solution, 50x is a chemstock used to prepare Bold Modified Freshwater Nutrient Solution for use in phycology/algae studies.

Formula variant

With major salts and trace elements. Modified with trace elements as requested for ASTM TOXICITY TESTS E1193 and E1295 conducted with Daphnia species.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karina M Michetti et al.
Biocell : official journal of the Sociedades Latinoamericanas de Microscopia Electronica ... et. al, 30(3), 491-496 (2007-03-23)
Nonspecific acid phosphatases are a group of enzymes whose activity increases the availability of exogenous and endogenous orthophosphate either through extra- or intracellular hydrolysis of phosphate compounds. Our study demonstrates the activity of acid phosphatases in the filamentous freshwater alga
Osamu Nishimura et al.
PloS one, 10(11), e0143525-e0143525 (2015-11-21)
We established a laboratory clonal strain of freshwater planarian (Dugesia japonica) that was derived from a single individual and that continued to undergo autotomous asexual reproduction for more than 20 years, and we performed large-scale genome sequencing and transcriptome analysis
Jakub Nedbal et al.
HardwareX, 8, e00143-e00143 (2021-01-15)
A bottom-illuminated orbital shaker designed for the cultivation of microalgae suspensions is described in this open-source hardware report. The instrument agitates and illuminates microalgae suspensions grown inside flasks. It was optimized for low production cost, simplicity, low power consumption, design
Sing-Lai Lim et al.
Bioresource technology, 101(19), 7314-7322 (2010-06-16)
The potential application of Chlorella vulgaris UMACC 001 for bioremediation of textile wastewater (TW) was investigated using four batches of cultures in high rate algae ponds (HRAP) containing textile dye (Supranol Red 3BW) or TW. The biomass attained ranged from
Eloísa Ramos-Rodríguez et al.
The Journal of experimental biology, 223(Pt 7) (2020-02-29)
Organisms with wide environmentally induced morphological plasticity and cosmopolitan distribution, e.g. the common freshwater rotifer Keratella cochlearis, are ideal models to study the evolution of plastic polymorphisms and the capacity of zooplankton to adapt to local selection conditions. We investigated

Protocols

Phycology media can be prepared from concentrated solutions or from powdered salt mixtures. The concentrated solutions are complete, including vitamins, and should be stored frozen.

Phycology media can be prepared from concentrated solutions or from powdered salt mixtures. The concentrated solutions are complete, including vitamins, and should be stored frozen.

Phycology media can be prepared from concentrated solutions or from powdered salt mixtures. The concentrated solutions are complete, including vitamins, and should be stored frozen.

Phycology media can be prepared from concentrated solutions or from powdered salt mixtures. The concentrated solutions are complete, including vitamins, and should be stored frozen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service