Skip to Content
Merck

B2891

BES sodium salt

≥99% (titration)

Synonym(s):

N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H14NNaO5S
CAS Number:
Molecular Weight:
235.23
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161700
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (titration)

form

crystalline powder

useful pH range

6.4-7.8

pKa (25 °C)

7.1 (Free acid)

solubility

water: 333.3 mg/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

[Na+].OCCN(CCO)CCS([O-])(=O)=O

InChI

1S/C6H15NO5S.Na/c8-4-1-7(2-5-9)3-6-13(10,11)12;/h8-9H,1-6H2,(H,10,11,12);/q;+1/p-1

InChI key

CFQLQLSIZOWFNV-UHFFFAOYSA-M



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library