Skip to Content
Merck

Skip To

B2805

BMP6, human

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

Synonym(s):

Bone Morphogenetic Protein 6 human, BMP-6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Biological source:
human
Recombinant:
expressed in NSO cells
Assay:
>95% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
30–38 kDa
Impurities:
endotoxin, tested
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Bone Morphogenetic Protein 6 human, >95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

biological source

human

Quality Segment

recombinant

expressed in NSO cells

assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

0.05-0.15 μg/mL ED50

mol wt

30–38 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... BMP6(654)

Biochem/physiol Actions

Cellular responses to BMP-6 are mediated by the formation of hetero-oligomeric complexes of type I and type II serine/threonine kinase receptors. Mature human and murine BMP-6 demonstrates 96% homology.

Physical form

Lyophilized from 200 ul 0.2 μm filtered solution in 40% acetonitrile, 0.1% trifluoroacetic acid containing 50ug bovine serum albumin per 1 ug as a carrier protein.

Analysis Note

The biological activity is measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B2680B3795B1434
assay

>95% (SDS-PAGE)

assay

>95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

>95% (SDS-PAGE)

biological source

human

biological source

human

biological source

human

biological source

human

recombinant

expressed in NSO cells

recombinant

expressed in NSO cells

recombinant

expressed in E. coli

recombinant

expressed in CHO cells

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters