Skip to Content
Merck
All Photos(1)

Key Documents

B2292

Sigma-Aldrich

O6-Benzylguanine

≥98% (TLC), solid, O⁶-alkylguanine DNA alkyltransferase inhiitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5O
CAS Number:
Molecular Weight:
241.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

O6-Benzylguanine, ≥98% (TLC), solid

Quality Level

Assay

≥98% (TLC)

form

solid

solubility

methanol: 20 mg/mL

storage temp.

room temp

SMILES string

Nc1nc(OCc2ccccc2)c3nc[nH]c3n1

InChI

1S/C12H11N5O/c13-12-16-10-9(14-7-15-10)11(17-12)18-6-8-4-2-1-3-5-8/h1-5,7H,6H2,(H3,13,14,15,16,17)

InChI key

KRWMERLEINMZFT-UHFFFAOYSA-N

Gene Information

human ... MGMT(4255)

Looking for similar products? Visit Product Comparison Guide

Application

O6-Benzylguanine has been used:
  • as an inhibitor of methylguanine methyltransferase (MGMT) in glioblastoma stem cell
  • as a O6-alkylguanine-alkyltransferase (AGT) enzyme inhibitor in embryonic stem cells prior to N-ethyl-N-nitrosourea(ENU) treatment
  • as an inhibitor of AGT in growth inhibition assays of HL-60 human promyelocytic leukemia cells

Biochem/physiol Actions

O(6)-benzylguanine is an antineoplastic agent that binds the DNA repair enzyme O(6)-alkylguanine DNA alkyltransferase (AGT), resulting in inhibition of AGT-mediated DNA repair. It is widely used in various DNA repair mechanism studies and potentiates the effects of other chemotherapeutic agents that damage DNA.
O6-Benzylguanine (O6BG) inhibits methylguanine methyltransferase (MGMT) by blocking the active site through benzyl group transfer. The use of O6BG with bis-chloroethylnitrosourea (BCNU) or carmustine is effective in treating solid tumors including lymphomas, melanomas and sarcoma.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lynn Martin et al.
Radiation research, 172(4), 405-413 (2009-09-24)
Low-dose hyper-radiosensitivity (HRS) is the phenomenon whereby cells exposed to radiation doses of less than approximately 0.5 Gy exhibit increased cell killing relative to that predicted from back-extrapolating high-dose survival data using a linear-quadratic model. While the exact mechanism remains
A Phase III study of radiation therapy (RT) and O 6-benzylguanine+ BCNU versus RT and BCNU alone and methylation status in newly diagnosed glioblastoma and gliosarcoma: Southwest Oncology Group (SWOG) study S0001
Blumenthal DT, et al.
International Journal of Clinical Oncology, 20(4), 650-658 (2015)
Brian C Beard et al.
The Journal of clinical investigation, 120(7), 2345-2354 (2010-06-17)
HSC transplantation using genetically modified autologous cells is a promising therapeutic strategy for various genetic diseases, cancer, and HIV. However, for many of these conditions, the current efficiency of gene transfer to HSCs is not sufficient for clinical use. The
Mako Kamiya et al.
Analytical chemistry, 82(15), 6472-6479 (2010-07-02)
We introduce here a new class of BODIPY-based Ca2+ indicators which can be derivatized with biological ligands that permit the localization of the indicators in living cells. The underivatized BODIPY-based Ca2+ indicator (BOCA-1) shows a 250-fold increase in fluorescence intensity
Katharina Stöhr et al.
Analytical chemistry, 82(19), 8186-8193 (2010-09-08)
Recent developments in fluorescence microscopy raise the demands for bright and photostable fluorescent tags for specific and background free labeling in living cells. Aside from fluorescent proteins and other tagging methods, labeling of SNAP-tagged proteins has become available thereby increasing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service