Skip to Content
Merck
All Photos(1)

Key Documents

B2134

Sigma-Aldrich

2-Bromo-α-ergocryptine methanesulfonate salt

solid

Synonym(s):

(+)-2-Bromo-12′-hydroxy-2′-(1-methylethyl)-5′-(2-methylpropyl)ergotaman-3′,6′-18-trione methanesulfonate salt, (+)-Bromocriptine methanesulfonate salt, Bromocriptine mesylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40BrN5O5 · CH4SO3
CAS Number:
Molecular Weight:
750.70
Beilstein:
4115238
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

optical activity

[α]20/D +95°, c = 1 in methanol: methylene chloride (1:1)(lit.)

color

white

solubility

H2O: 0.8 mg/mL
ethanol: 23 mg/mL

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.[H][C@@]12Cc3c(Br)[nH]c4cccc(C1=CC(CN2C)C(=O)N[C@@]5(O[C@]6(O)N([C@@H](CC(C)C)C(=O)N7CCC[C@@]67[H])C5=O)C(C)C)c34

InChI

1S/C32H40BrN5O5.CH4O3S/c1-16(2)12-24-29(40)37-11-7-10-25(37)32(42)38(24)30(41)31(43-32,17(3)4)35-28(39)18-13-20-19-8-6-9-22-26(19)21(27(33)34-22)14-23(20)36(5)15-18;1-5(2,3)4/h6,8-9,13,16-18,23-25,34,42H,7,10-12,14-15H2,1-5H3,(H,35,39);1H3,(H,2,3,4)/t18?,23-,24+,25+,31-,32+;/m1./s1

InChI key

NOJMTMIRQRDZMT-NEKRQHSLSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromo-α-ergocryptine methanesulfonate salt has been used:
  • as D2 agonist in bird zebra finches
  • for Prl secretion inhibitor in mice
  • for the inhibition of motility in planaria worm

Biochem/physiol Actions

Bromocriptine is an ergot alkaloid and a dopamine D2 receptor agonist. It is prescribed for Parkinson′s disorder, hyperprolactinemia and galactorrhoea. It modulates β cells of the pancreas from insulin hypersecretion and improves the metabolic profile in type 2 diabetes patients with obesity. It also modulates glutamate release by glutamate transporter,GLT-1.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bromocriptine: old drug, new formulation and new indication.
Holt RI, et al.
Diabetes, obesity & metabolism, 12(12), 1048-1057 (2010)
S Perachon et al.
European journal of pharmacology, 366(2-3), 293-300 (1999-03-19)
We measured the affinities of bromocriptine, pramipexole, pergolide and ropinirole at human recombinant dopamine D1, D2 and D3 receptors in binding and functional tests. All four compounds bound with high affinity at the dopamine D3 receptor; bromocriptine and pergolide also
Prolactin and glucocorticoid signaling induces lactation-specific tight junctions concurrent with beta-casein expression in mammary epithelial cells.
Kobayashi K, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1863(8), 2006-2016 (2016)
Bromocriptine, an ergot alkaloid, inhibits excitatory amino acid release mediated by glutamate transporter reversal.
Shirasaki Y, et al.
European Journal of Pharmacology, 643(1), 48-57 (2010)
Lowering prolactin reduces post-hatch parental care in male and female zebra finches (Taeniopygia guttata).
Smiley KO and Adkins-Regan E
Hormones and Behavior, 98(1), 103-114 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service