Skip to Content
Merck

B169

BRL 37344 sodium salt hydrate

≥98% (HPLC), β3-adrenergic receptor agonist, solid

Synonym(s):

(±)-(R*,R*)-[4-[2-[[2-(3-Chlorophenyl)-2-hydroxyethyl]amino]propyl]phenoxy]acetic acid sodium hydrate, BRL 37344A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H21ClNNaO4 · xH2O
CAS Number:
Molecular Weight:
385.82 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

BRL 37344 sodium salt hydrate, solid

assay

≥98% (HPLC)

Quality Segment

form

solid

color

white

solubility

DMSO: 18 mg/mL, clear (warmed)

SMILES string

O.[Na+].C[C@H](Cc1ccc(OCC([O-])=O)cc1)NC[C@H](O)c2cccc(Cl)c2

InChI

1S/C19H22ClNO4.Na.H2O/c1-13(21-11-18(22)15-3-2-4-16(20)10-15)9-14-5-7-17(8-6-14)25-12-19(23)24;;/h2-8,10,13,18,21-22H,9,11-12H2,1H3,(H,23,24);;1H2/q;+1;/p-1/t13-,18+;;/m1../s1

InChI key

YTPAKDODIXRQTF-BNTHFPBMSA-M

Gene Information

human ... ADRB3(155)

Biochem/physiol Actions

BRL 37344 is a selective β3-adrenoceptor agonist.
BRL 37344 is a selective β3-adrenoceptor agonist. Studies support the hypothesis that there are peripheral β3 adrenergic receptors that can reduce food intake and that central β2 or β3 adrenergic receptors mediate the peripheral effects of the β3 agonist.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Disclaimer

Hygroscopic; store desiccated.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
B169-5MG04061832088815
B169-25MG04061833426326