Skip to Content
Merck
All Photos(1)

Key Documents

B133

Sigma-Aldrich

(S)-(−)-Bay K8644

≥98% (HPLC), solid

Synonym(s):

S-(−)-1,4-Dihydro-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-3-pyridinecarboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15F3N2O4
CAS Number:
Molecular Weight:
356.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

H2O: slightly soluble
ethanol: soluble (Organic solutions are stable for at least 20 days at RT.)

storage temp.

2-8°C

SMILES string

COC(=O)C1=C(C)NC(C)=C([C@H]1c2ccccc2C(F)(F)F)[N+]([O-])=O

InChI

1S/C16H15F3N2O4/c1-8-12(15(22)25-3)13(14(21(23)24)9(2)20-8)10-6-4-5-7-11(10)16(17,18)19/h4-7,13,20H,1-3H3/t13-/m0/s1

InChI key

ZFLWDHHVRRZMEI-ZDUSSCGKSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-Bay K8644, a dihydropyridine compound is a Ca2+ channel activator. It is an enantiomer of Bay K 8644.

Application

(S)-(-)-Bay K8644 has been used as a L-type calcium channel (LTCC) agonist in the suprachiasmatic nucleus (SCN) slices. It has also been used as a LTCC agonist to induce arrhythmia in rats.

Biochem/physiol Actions

(S)-(-)-Bay K8644 enhances Ca2+ current, whereas the (+)-Bay K 8644 is a weak Ca2+ channel antagonist. Bay K 8644 is lipophilic and it effectively scavenges superoxide anions.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Manufactured and sold with the permission of Bayer AG.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua P Whitt et al.
The Journal of general physiology, 150(2), 259-275 (2017-12-15)
Large conductance K+ (BK) channels are expressed widely in neurons, where their activation is regulated by membrane depolarization and intracellular Ca2+ (Ca2+i). To enable this regulation, BK channels functionally couple to both voltage-gated Ca2+ channels (VGCCs) and channels mediating Ca2+
Gildenisson Araújo do Nascimento et al.
Arquivos brasileiros de cardiologia, 113(5), 925-932 (2019-09-05)
D-limonene (DL) is a monoterpene and is the major component in the essential oil of citrus fruit. It presents antihyperglycemic and vasodilatation activities. This study evaluated the cardiovascular effects and potential antiarrhythmic of DL in rats. Hemodynamic and electrocardiographic (ECG)
J J Enyeart et al.
Molecular endocrinology (Baltimore, Md.), 4(5), 727-735 (1990-05-01)
Dihydropyridine (DHP) Ca2+ channel modulators were used to explore the relationship between voltage-gated Ca2+ channels and PRL secretion, synthesis, and mRNA in PRL-secreting pituitary cells. Optical isomers of the Ca2+ channel agonist Bay K 8644 produced stereospecific and opposing effects
J Ferrante et al.
Biochemical and biophysical research communications, 158(1), 149-154 (1989-01-16)
The 1,4-dihydropyridine Ca2+ channel activator, (-) [3H]Bay K 8644, binds to cardiac membranes and polarized [5 mM K+] and depolarized [50 mM K+] cardiac cells. Binding to microsomal membranes at 25 degrees C indicates a single set of binding sites
Herbert Marini et al.
Frontiers in endocrinology, 3, 94-94 (2012-10-13)
Genistein aglycone, one of the soy isoflavones, has been reported to be beneficial in the treatment of menopausal vasomotor symptoms, osteoporosis, and cardiovascular diseases, as well as in a variety of cancers. However, issues of potential harm on thyroid function

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service