Skip to Content
Merck
All Photos(2)

Key Documents

B1022

Sigma-Aldrich

Biotinamidohexanoic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt

≥90% (TLC), powder

Synonym(s):

Sulfosuccinimidyl 6-(biotinamido)hexanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29N4NaO9S2
CAS Number:
Molecular Weight:
556.59
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.46

Assay

≥90% (TLC)

form

powder

solubility

H2O: 10 mg/mL
DMF: 50 mg/mL

storage temp.

−20°C

SMILES string

[Na].OS(=O)(=O)C1CC(=O)N(OC(=O)CCCCCNC(=O)CCCCC2SCC3NC(=O)NC23)C1=O

InChI

1S/C20H30N4O9S2.Na.H/c25-15(7-4-3-6-13-18-12(11-34-13)22-20(29)23-18)21-9-5-1-2-8-17(27)33-24-16(26)10-14(19(24)28)35(30,31)32;;/h12-14,18H,1-11H2,(H,21,25)(H2,22,23,29)(H,30,31,32);;

InChI key

KDRVGWAQYRWAJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Biotinamidohexanoic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt is a water soluble biotinylation reagent. It can be used in immunoprecipitation of signaling proteins. It can also be used in analysis of hyaluronan synthesis. Further, the product can also be used in ELISA. This biotinylation reagent has shown specificity for human dermal fibroblasts.

Application

Water soluble biotinylation reagent incorporating an aminocaproyl spacer. This can reduce the steric hindrance in binding avidin to some biotinylated compounds. Has also been used as a cell surface labeling reagent. Typically coupled to primary amine in the pH range 6.5-8.5. May be used for the biotinylation of cell surface proteins.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Schäfer-Somi et al.
Reproduction in domestic animals = Zuchthygiene, 41(1), 48-54 (2006-01-20)
Sera of healthy pregnant (group I, n = 11) and non-pregnant (group II, n = 11) bitches were screened for autoantibodies (AAb). In both groups, blood samples were drawn every fifth day between days 5 and 55 after mating. Serum
A V Vener et al.
Analytical biochemistry, 191(1), 65-69 (1990-11-15)
A colorimetric procedure to detect biotin bound to microtiter plates with a sensitivity down to 10(-16) mol was developed using biotinylated inorganic pyrophosphatase of Escherichia coli. Reaction of pyrophosphatase with 1 mM N-biotinyl-6-aminocaproic acid N-hydroxy-sulfonosuccinimide ester yielded a stable 87%
H Watarai et al.
Electrophoresis, 21(2), 460-464 (2000-02-16)
The expression of plasma membrane proteins in human monocyte-derived U937 cells was examined by cell disruption and isolation of microsomal fractions. Two alternative procedures for cell disruption, Dounce homogenization and nitrogen cavitation, were compared. Cell homogenization and sequential centrifugation resulted
Improved nonradioactive cell surface labeling technique for immunoprecipitation.
L M Lantz et al.
BioTechniques, 18(1), 56-56 (1995-01-01)
Mohammed M Nooh et al.
American journal of physiology. Renal physiology, 317(2), F375-F387 (2019-05-30)
Arginine-vasopressin (AVP)-mediated translocation of aquaporin-2 (AQP2) protein-forming water channels from storage vesicles to the membrane of renal collecting ducts is critical for the renal conservation of water. The type-1 PDZ-binding motif (PBM) in AQP2, "GTKA," is a critical barcode for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service