Skip to Content
Merck

B016

Benoxathian hydrochloride

solid

Synonym(s):

2-[[[2-(2,6-Dimethoxyphenoxy)ethyl]-amino]-methyl]-1,4-benzoxathian hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H23NO4S · HCl
CAS Number:
Molecular Weight:
397.92
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

color

white

solubility

H2O: soluble (Aqueous solutions may be stored for several weeks at -20 °C.), ethanol: soluble, isopropanol: soluble

SMILES string

Cl.COc1cccc(OC)c1OCCNCC2CSc3ccccc3O2

InChI

1S/C19H23NO4S.ClH/c1-21-16-7-5-8-17(22-2)19(16)23-11-10-20-12-14-13-25-18-9-4-3-6-15(18)24-14;/h3-9,14,20H,10-13H2,1-2H3;1H

InChI key

OWRADFDDJVNMGL-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Selective α1-adrenoceptor antagonist.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library