Skip to Content
Merck

AXYAM2MLRD

Corning® Axygen® AxyMats® 96 Round Well Sealing Mat for Deep Well Plate

for 2 mL round deep well plates, size 96 wells, pkg of 5x10mats/cs

Synonym(s):

Sealing mat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122410
Manufacturer/tradename:
Axygen Inc AM-2ML-RD, Corning
Material:
silicone (research-grade)
Sterility:
non-sterile
Packaging:
pkg of 5x10mats/cs
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

silicone (research-grade)

sterility

non-sterile

manufacturer/tradename

Axygen Inc AM-2ML-RD, Corning

packaging

pkg of 5x10mats/cs

size

96 wells

Application

Corning® Axygen® AxyMats® 96 Round Well Sealing Mat for Deep Well Plate has been used in genomic DNA extraction from Sporosarcina pasteurii.

Features and Benefits

  • Made from research-grade silicone
  • Follows ethanol rinse and bleach wash protocol for reuse
  • Reduce evaporation in PCR and storage applications over a wide range of temperatures
  • Compatible with P-DW-20-C, P-96-450V-C, P-DW-11-C, P-96-450R-C, assay,96 well deep well, and storage plates
  • Autoclavable

Legal Information

AxyMats is a registered trademark of Corning, Inc.
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library