Skip to Content
Merck
All Photos(5)

Key Documents

AV44108

Sigma-Aldrich

Anti-SLC15A4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FP12591, Anti-PHT1, Anti-PTR4, Anti-Solute carrier family 15, member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC15A4

Application

Anti-SLC15A4 antibody produced in rabbit is suitable for western blotting at a concentration of 0.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

SLC15A4 (PHT1; PTR4) is a peptide/histidine transporter expressed in the brain and the retina. This protein enables plasmacytoid dendritic cells to sense toll-like receptor ligands that aid in triggering immune responses to persistent viral infections. The expression of SLC15A4 has been implicated in the pathogenesis of lupus.

Sequence

Synthetic peptide located within the following region: MEGSGGGAGERAPLLGARRAAAAAAAAGAFAGRRAACGAVLLTELLERAA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Baccala et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(8), 2940-2945 (2013-02-06)
In vitro evidence suggests that plasmacytoid dendritic cells (pDCs) are intimately involved in the pathogenesis of lupus. However, it remains to be determined whether these cells are required in vivo for disease development, and whether their contribution is restricted to
Amanda L Blasius et al.
PLoS pathogens, 8(9), e1002915-e1002915 (2012-10-03)
Plasmacytoid dendritic cells (pDCs) are the major producers of type I IFN in response to viral infection and have been shown to direct both innate and adaptive immune responses in vitro. However, in vivo evidence for their role in viral
T Yamashita et al.
The Journal of biological chemistry, 272(15), 10205-10211 (1997-04-11)
Here we report the cloning and functional characterization of a rat novel peptide/histidine transporter (PHT1), which was expressed in the brain and the retina. The cDNA encodes the predicted protein of 572 amino acid residues with 12 putative membrane-spanning domains.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service