Skip to Content
Merck
All Photos(3)

Key Documents

AV38692

Sigma-Aldrich

Anti-SMAD1 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-BSP1, Anti-JV4-1, Anti-JV41, Anti-MADH1, Anti-MADR1, Anti-SMAD family member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

rat, mouse, human, dog, rabbit, guinea pig, bovine, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD1(4086)

Immunogen

Synthetic peptide directed towards the N terminal region of human SMAD1

Biochem/physiol Actions

SMAD proteins are transcription regulators that mediate multiple signal transduction pathways. They are involved in a wide variety of cellular processes such as cell growth, apoptosis, survival, differentiation, morphogenesis and immune responses. SMAD1 interacts with PAWS1/FAM83G protein and regulates bone morphogenetic protein signaling.

Sequence

Synthetic peptide located within the following region: MNVTSLFSFTSPAVKRLLGWKQGDEEEKWAEKAVDALVKKLKKKKGAMEE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshio Matsumoto et al.
Endocrine journal, 59(2), 91-101 (2011-09-21)
Mechanical stress and parathyroid hormone (PTH) are major stimulators, and aging and glucocorticoids excess are important suppressors of osteoblast differentiation. Mechanical stress and PTH stimulate interleukin (IL)-11 expression in cells of osteoblast lineage by enhancing transcription of IL-11 gene via
Janis Vogt et al.
Open biology, 4, 130210-130210 (2014-02-21)
Bone morphogenetic proteins (BMPs) control multiple cellular processes in embryos and adult tissues. BMPs signal through the activation of type I BMP receptor kinases, which then phosphorylate SMADs 1/5/8. In the canonical pathway, this triggers the association of these SMADs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service