Skip to Content
Merck
All Photos(2)

Documents

AV38611

Sigma-Aldrich

Anti-MyB antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cmyb, Anti-Efg, Anti-c-myb, Anti-c-myb_CDS, Anti-v-Myb myeloblastosis viral oncogene homolog (avian)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

mouse, bovine, human, horse, dog, rabbit, guinea pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYB(4602)

Immunogen

Synthetic peptide directed towards the N terminal region of human MYB

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

MYB may be a transcriptional activator; DNA-binding protein that specifically recognize the sequence 5′-YAAC[GT]G-3′. It plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.

Sequence

Synthetic peptide located within the following region: YDGLLPKSGKRHLGKTRWTREEDEKLKKLVEQNGTDDWKVIANYLPNRTD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingtao Na et al.
Cell & bioscience, 11(1), 31-31 (2021-02-08)
Heart failure (HF) is a clinical syndrome characterized by left ventricular dysfunction or elevated intracardiac pressures. Research supports that microRNAs (miRs) participate in HF by regulating  targeted genes. Hence, the current study set out to study the role of HDAC3-medaited
Petr Kaspar et al.
PloS one, 8(10), e76742-e76742 (2013-11-10)
Satellite cells represent a heterogeneous population of stem and progenitor cells responsible for muscle growth, repair and regeneration. We investigated whether c-Myb could play a role in satellite cell biology because our previous results using satellite cell-derived mouse myoblast cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service