Skip to Content
Merck
All Photos(2)

Key Documents

APOAF

Sigma-Aldrich

Annexin V-FITC Apoptosis Detection Kit

Synonym(s):

Annexin V-FITC, Apoptosis probe FITC

Sign Into View Organizational & Contract Pricing

Select a Size

20 TEST
€329.00
60 TEST
€707.00

€329.00


In StockDetails


To order products, please contact your local dealer.

Select a Size

Change View
20 TEST
€329.00
60 TEST
€707.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.84

€329.00


In StockDetails


To order products, please contact your local dealer.

usage

(20 tests)

Quality Level

packaging

pkg of 1 kit

technique(s)

flow cytometry: suitable

application(s)

cell analysis
detection

detection method

fluorometric

shipped in

wet ice

storage temp.

2-8°C

General description

Annexin V-FITC kit allows fluorescent detection of annexin V bound to apoptotic cells and quantitative determination by flow cytometry. The AnnexinV-FITC kit uses annexin V conjugated with fluorescein isothiocyante (FITC) to label phosphatidylserine sites on the membrane surface. The kit includes propidium iodide (PI) to label the cellular DNA in necrotic cells where the cell membrane has been totally compromised. This combination allows the differentiation among early apoptotic cells (annexin V positive, PI negative), necrotic cells (annexin V positive, PI positive), and viable cells (annexin V negative, PI negative).

Application

Annexin V-FITC Apoptosis Detection Kit was used:
  • in staining of LNCaP prostate cancer cells for measuring the G. lucidum extracts activity during the treatment of prostate cancer.[1]
  • for tumor cell labelling to study the inhibitory activity of DBP-maf (Vitamin D binding protein-macrophage activating factor) on prostate tumor cells.[2]
  • for indirect measurement of flippase activity.[3]

Features and Benefits

Detects apoptosis earlier in the process than DNA-based assays such as TUNEL.
  • Rapid labeling of cells. Cell staining takes only 10 minutes.
  • No cell fixation or processing required, reducing the detection time and allowing the cells to be used for further study.
  • Propidium iodide secondary dye is included with the kit to differentiate apoptotic cells from viable and necrotic cells.

Other Notes

Allow all components to reach room temperature before use.

Kit Components Only

Product No.
Description

  • APOAFA

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise C Arruda et al.
The Journal of biological chemistry, 287(18), 14912-14922 (2012-02-16)
Complementarity-determining regions (CDRs) from monoclonal antibodies tested as synthetic peptides display anti-infective and antitumor activities, independent of the specificity of the native antibody. Previously, we have shown that the synthetic peptide C7H2, based on the heavy chain CDR 2 from
Monika Kallubai et al.
Journal of biomolecular structure & dynamics, 37(3), 623-640 (2018-01-30)
Our study focus on the biological importance of synthesized 5β-dihydrocortisol (Dhc) and 5β-dihydrocortisol acetate (DhcA) molecules, the cytotoxic study was performed on breast cancer cell line (MCF-7) normal human embryonic kidney cell line (HEK293), the IC50 values for MCF-7 cells
C Pigault et al.
Journal of molecular biology, 236(1), 199-208 (1994-02-11)
Annexins are intracellular proteins which bind to membranes in a Ca(2+)-dependent manner and which have been proposed to play regulatory roles in different membrane processes. In the present study, the stoichiometry of the Ca(2+)-dependent binding of annexin V to phosphatidylserine
Jianzhong Li et al.
Scientific reports, 6, 23975-23975 (2016-04-08)
Metformin, one of the most common prescriptions for patients with type 2 diabetes, is reported to protect the kidney from gentamicin-induced nephrotoxicity. However, the role and mechanisms for metformin in preventing cisplatin-induced nephrotoxicity remains largely unknown. In this study, a
Kalvin J Gregory et al.
PloS one, 5(10), e13428-e13428 (2010-10-27)
Vitamin D binding protein-macrophage activating factor (DBP-maf) is a potent inhibitor of tumor growth. Its activity, however, has been attributed to indirect mechanisms such as boosting the immune response by activating macrophages and inhibiting the blood vessel growth necessary for

Articles

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

See All

Questions

1–6 of 6 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Can Product APOAF, Annexin V-FITC Apoptosis Detection Kit be used on fixed cells?

    1 answer
    1. No. Product APOAF, Annexin V-FITC Apoptosis Detection Kit must be performed on live cells in order to measure Apoptosis. The assay is based on the externization of phosphatidylserine from the inner cell membrane to the outer cell membrane.  If the membrane is preturbed due to fixation, non-speciifc staining of the inner cell membrane might occur

      Helpful?

  3. When using Product APOAF, Annexin V-FITC Apoptosis Detection Kit, I have cells that are Annexin V FITC negative and PI positive.  What are these cells?

    1 answer
    1. It is not possible to have cells that are PI positive without the cells also being positive for Annexin V binding.  Cells are PI positive because the membrane has been compromised.  If this is the case, Annexin V can also enter the cell and bind to the PS on the internal cell membrane. The gating on the histogram for the FITC channel should be changed so that all cells that are PI positive are also Annexin V positive.

      Helpful?

  4. When using the Annexin V-FITC Apoptosis Detection Kit, Product APOAF, can I use any buffer for resuspending my cells?

    1 answer
    1. The binding buffer included in the kit needs to be used for resuspending cells.  The buffer contains calcium chloride at a final (1X) concentration of 2.5 mM which is necessary for the binding of annexin V to phosphatidylserine.

      Helpful?

  5. What wavelength do I use to detect Annexin V-FITC and Propidium Iodide when using Annexin V-FITC Apoptosis Detection Kit, Product APOAF?

    1 answer
    1. Annexin V FITC will have a maximum emission of 528 nm. This can be measured in the standard FITC Channel on a flow cytometer (FL1).  Propidium Iodide has a maximum emission of 620 nm.  This is measured on the short red channel on a flow cytometer (FL2 or FL3).

      Helpful?

  6. Can I use Product APOAF, Annexin V-FITC Apoptosis Detection Kit, to differentiate cells that are dead due to necrosis or apoptosis?

    1 answer
    1. When using a kinetic study (various time points), you can show the progression of the cells from viable (annexin V FITC negative, PI negative), to annexin FITC positive, PI negative (membrane flip) to annexin V FITC positive, PI positive (dead).  If there are cells that are double positive when starting, it is not possible to guarantee that the cell death occurred due to apoptosis with this assay.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service