Skip to Content
Merck

AF1

Aflatoxin kits

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41100000
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

2-8°C

Quality Level

Packaging

Four aflatoxins from Aspergillus flavus


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Aflatoxin B1 1 mg
    SDS

  • Aflatoxin B2 1 mg
    SDS

  • Aflatoxin G1 1 mg
    SDS

  • Aflatoxin G2 1 mg
    SDS

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Carc. 1B - Muta. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sven C Neulinger et al.
Applied and environmental microbiology, 75(5), 1437-1444 (2008-12-31)
The cold-water coral Lophelia pertusa (Scleractinia, Caryophylliidae) is a key species in the formation of cold-water reefs, which are among the most diverse deep-sea ecosystems. It occurs in two color varieties: white and red. Bacterial communities associated with Lophelia have



Global Trade Item Number

SKUGTIN
0710-264904065272564786
0710-356504065273456721
0710-332804065273456691
0710-318204065273456622
0710-067104065272564403
0710-014004065272564113
0710-020404065272564175
AF1-1KT04061833418420