Skip to Content

A9325

L-Alanine 4-nitroanilide hydrochloride

protease substrate, ≥99% (TLC), powder

Synonym(s):

Ala-4-nitroanilide, H-Ala-pNA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H11N3O3 · HCl
CAS Number:
Molecular Weight:
245.66
NACRES:
NA.83
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352204
EC Number:
250-811-0
MDL number:
Beilstein/REAXYS Number:
6303731
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Alanine 4-nitroanilide hydrochloride, protease substrate

Quality Segment

assay

≥99% (TLC)

form

powder

solubility

water: 50 mg/mL, clear to very slightly hazy

storage temp.

−20°C

SMILES string

Cl[H].C[C@H](N)C(=O)Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C9H11N3O3.ClH/c1-6(10)9(13)11-7-2-4-8(5-3-7)12(14)15;/h2-6H,10H2,1H3,(H,11,13);1H/t6-;/m0./s1

InChI key

YEXRLSXNWLNHQR-RGMNGODLSA-N

Application

L-Alanine 4-nitroanilide hydrochloride has been used to determine the activity of aminopeptidase in the intestinal segments and small intestine (jejunum and ileum).

Biochem/physiol Actions

L-Alanine-4-nitroanilide is used as a substrate to determine alanine aminopeptidase (AAP) activity. It is mainly chosen as a substrate, due to its relative specificity, higher hydrolysis rate and better solubility.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library