Skip to Content
Merck
All Photos(2)

Key Documents

A9223

ART® self-sealing barrier pipette tips

ART 10F, volume range 0.5-10 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NB.01
volume:
0.5—10 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
case of 5 packs
pack of 960 ea (Racked)
manufacturer/tradename:
Molecular BioProducts 2139F

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

case of 5 packs
pack of 960 ea (Racked)

manufacturer/tradename

Molecular BioProducts 2139F

technique(s)

PCR: suitable

maximum volume

10 μL

volume range

0.5-10 μL

volume

0.5—10 μL

compatibility

for use with

  • Biohit Variable .5-10 μl
  • Eppendorf Research Pro 4860 5-100 μl
  • Eppendorf Titerman 4908 5-50 μl
  • Finnpipette 4027 .5-10 μl
  • Finnpipette 4500 Digital .5-10 μl

Looking for similar products? Visit Product Comparison Guide

General description

Features the MicroPoint design for the Finnpipette

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)
Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service