Skip to Content
Merck
All Photos(3)

Documents

A9043

Sigma-Aldrich

S-Acetylthioglycolic acid N-hydroxysuccinimide ester

≥95% (TLC), powder

Synonym(s):

N-Succinimidyl (acetylthio)acetate, N-Succinimidyl S-acetylthioglycolate, S-Acetylthioglycolic acid NHS ester, SATA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO5S
CAS Number:
Molecular Weight:
231.23
Beilstein:
7815491
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95% (TLC)

form

powder

solubility

acetonitrile: 50 mg/mL
DMF: soluble

storage temp.

−20°C

SMILES string

CC(=O)SCC(=O)ON1C(=O)CCC1=O

InChI

1S/C8H9NO5S/c1-5(10)15-4-8(13)14-9-6(11)2-3-7(9)12/h2-4H2,1H3

InChI key

FLCQLSRLQIPNLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thiolating reagent for primary amines; thiol can be deprotected under mild conditions. Typically, couples initially to a molecule containing primary amine by amide bond buffered at pH 7.5. Deprotection may be accomplished with 0.05 M hydroxylamine at neutral pH. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

Other Notes

Note that the ester couples very efficiently to primary amines compared to other thiolating reagents. In addition, this reagent will neutralize original amine positive charge.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Wang et al.
Nature protocols, 2(4), 972-978 (2007-04-21)
The mercaptoacetyltriglycine (MAG3) chelator has been shown to stably complex technetium-99m (99mTc) for nuclear imaging and radiorhenium (186/188Re) for tumor radiation therapy studies. The bifunctional N-hydroxysuccinimidyl ester of MAG3 with S-acetyl protection (N-hydroxysuccinimidyl S-acetylmercaptoacetyltriglycinate (NHS-MAG3)) has been successfully used to
Jing-Tang Lin et al.
PloS one, 7(4), e36086-e36086 (2012-05-09)
To develop a fluorescent ruthenium complex for biosensing, we synthesized a novel sulfhydryl-reactive compound, 4-bromophenanthroline bis-2,2'-dipyridine Ruthenium bis (hexafluorophosphate). The synthesized Ru(II) complex was crosslinked with thiol-modified protein G to form a universal reagent for fluorescent immunoassays. The resulting Ru(II)-protein
M H Vingerhoeds et al.
Pharmaceutical research, 13(4), 604-610 (1996-04-01)
Immuno-enzymosomes are tumor-specific immunoliposomes bearing enzymes on their surface. These enzymes are capable of converting relatively nontoxic prodrugs into active cytostatic agents. The enzyme beta-glucuronidase (GUS)4 was coupled to the external surface of immunoliposomes directed against ovarian carcinoma cells. This
R A Schwendener et al.
Biochimica et biophysica acta, 1026(1), 69-79 (1990-07-09)
The two coupling agents SPDP (N-succinimidyl-3-(2-pyridyldithio)propionate) and SATA (N-succinimidyl-S-acetylthioacetate) were compared in their efficiency and feasibility to couple monoclonal antibodies (Abs) via thioether linkage to liposomes functionalized by various lipophilic maleimide compounds like N-(3-maleimidopropionyl)-N2-palmitoyl-L-lysine methyl ester (MP-PL), N-(3-maleimidopropionyl)phosphatidylethanolamide (MP-PE), N6-(6-maleimidocaproyl)-N2-palmitoyl-L-lysine
Nejat Düzgüneş et al.
Methods in enzymology, 391, 351-373 (2005-02-22)
The intracellular activity of certain antiviral agents, including antisense oligonucleotides, acyclic nucleoside phosphonates, and protease inhibitors, is enhanced when they are delivered in liposome-encapsulated form. In this chapter we describe the preparation of pH-sensitive liposomes encapsulating antisense oligonucleotides, ribozymes, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service