Skip to Content
Merck

A8978

Anti-Beta (β)-Amyloid antibody

mouse monoclonal, BAM90.1

Synonym(s):

Anti-Aβ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
BAM90.1, monoclonal
Application:
EIA, IHC, IP, WB
Citations:
13
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-β-Amyloid (13-28) antibody, Mouse monoclonal, clone BAM90.1, purified from hybridoma cell culture

biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BAM90.1, monoclonal

form

buffered aqueous solution

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

enzyme immunoassay: 0.2-0.4 μg/mL using amyloid β-protein, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APP(351)

General description

Amyloids are insoluble protein aggregates consisting of misfolded proteins and peptides. Amyloid deposition are associated with multiple neurodegenerative disorders.
The β-amyloid precursor protein (APP) is cleaved sequentially by the proteolytic enzymes β-secretase (BACE1) and γ-secretase to produce β-amyloid (Aβ) peptides with the Aβ1-42 and the Aβ1-40 forms being the most prevalent. Secreted Aβ peptides are degraded either via a re-uptake mechanism followed by endosomal degradation, or by an extracellular insulin degrading enzyme. Extracellular accumulation of Aβ leads to the formation of aggregates, fibrils and eventually amyloid deposits called neuritic plaques, which is the hallmark of Alzheimer′s disease (AD).
Monoclonal Anti β-Amyloid [13-28] recognizes the β-Amyloid peptide. The antibody epitope resides within amino acids 20-23.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Mouse Monoclonal Anti-β-Amyloid (13-28) antibody can be used for immunoblotting assays at 1:5,000. The antibody can also be used for IHC, immunoprecipitation,fluorescence infrared spectroscopy and enzyme immunoassays (02-0.4μg/ml).

Physical form

Filtered solution in 0.01 M phosphate buffered saline, pH 7.4.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library