Skip to Content
Merck
All Photos(1)

Key Documents

A8843

Sigma-Aldrich

AM580

≥98% (HPLC)

Synonym(s):

4-[(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)carboxamido]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO3
CAS Number:
Molecular Weight:
351.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC1(C)CCC(C)(C)c2cc(ccc12)C(=O)Nc3ccc(cc3)C(O)=O

InChI

1S/C22H25NO3/c1-21(2)11-12-22(3,4)18-13-15(7-10-17(18)21)19(24)23-16-8-5-14(6-9-16)20(25)26/h5-10,13H,11-12H2,1-4H3,(H,23,24)(H,25,26)

InChI key

SZWKGOZKRMMLAJ-UHFFFAOYSA-N

Gene Information

Related Categories

General description

AM580 is a retinobenzoic derivative and a RAR-α (retinoic acid receptor-α) agonist. It stimulates the maturation of granulocytes in NB4 promyelocytic leukemia cell line and APL (acute promyelocytic leukemia) blasts. AM580 suppresses endometrial cancer cell proliferation.

Application

Retinoid derivative used for gene expression studies of leukemia-retinoic acid receptor (PHL-RAR) cell line differentiation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endocrinologia Experimentalis (2012)
Chunyue Yin et al.
Hepatology (Baltimore, Md.), 56(5), 1958-1970 (2012-04-11)
Hepatic stellate cells (HSCs) are liver-specific mesenchymal cells that play vital roles in liver development and injury. Our knowledge of HSC biology is limited by the paucity of in vivo data. HSCs and sinusoidal endothelial cells (SECs) reside in close
Vitamin A in Health and Disease (2007)
S C Brooks et al.
Blood, 87(1), 227-237 (1996-01-01)
The ability of subtypes of retinoic acid receptors (RARs) and retinoid X receptors (RXRs) singly and in combination to elicit myeloid differentiation, G1/0-specific growth arrest, and retinoblastoma (RB) tumor suppressor protein dephosphorylation was determined in the human myeloblastic leukemia cell
Synthesis and pharmacological characterization of Disila-AM80 (Disila-tamibarotene) and Disila-AM580, silicon analogues of the RARalpha-selective retinoid agonists AM80 (Tamibarotene) and AM580.
Reinhold Tacke et al.
ChemMedChem, 4(11), 1797-1802 (2009-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service