Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
bovine
Quality Segment
form
powder
mol wt
~66,000
packaging
vial of 50 mg
origin
USA origin
technique(s)
ELISA: suitable, gel permeation chromatography (GPC): suitable, immunohistochemistry: suitable
impurities
~0.3% dithiothreitol
pH
<5.0
suitability
suitable for (Suitable for use as a gel filtration marker)
UniProt accession no.
foreign activity
BT virus, none detected, VSV virus, none detected
storage temp.
−20°C
Gene Information
bovine ... ALB(280717)
General description
Application
- for blocking of nonspecific immunoglobulin during immunohistochemical reactions.[1]
- to coat microplates for blocking non-specific binding during I-ELISA (Indirect Enzyme-linked Immunosorbent Assay)
- as molecular mass standard in analytical gel filtration chromatography for Superdex® 75 10/300 GL gel filtration column.[2]
- for measuring fluorescence for PR1 (phytofluor red 1) by using Fluorescence correlation spectroscopy (FCS)[3]
- as a reference mixture along with horse Mb in Size Exclusion Chromatography[4]
Biochem/physiol Actions
Preparation Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| biological source bovine | biological source bovine | biological source bovine | biological source bovine |
| technique(s) ELISA: suitable, immunohistochemistry: suitable, gel permeation chromatography (GPC): suitable | technique(s) ELISA: suitable, blocking: suitable, electrophoresis: suitable, western blot: suitable | technique(s) blood typing: suitable, microbiological culture: suitable | technique(s) blood typing: suitable |
| origin USA origin | origin USA origin | origin USA origin | origin USA origin |
| Quality Level 200 | Quality Level 300 | Quality Level 200 | Quality Level 200 |
| suitability suitable for (Suitable for use as a gel filtration marker) | suitability - | suitability - | suitability - |
| form powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
Still not finding the right product?
Explore all of our products under Bovine Serum Albumin
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

