Skip to Content

System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.

Merck

Key Documents

Skip To

A8301

Albumin, glycated human

lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25
MDL number:
eCl@ss:
42010201
Biological source:
human
Assay:
90-100% protein basis (biuret)
Form:
lyophilized powder
Technique(s):
cell culture | mammalian: suitable
Concentration:
90—100% protein (Biuret)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

assay

90-100% protein basis (biuret)

form

lyophilized powder

concentration

90—100% protein (Biuret)

extent of labeling

1-5 mol hexose (as fructosamine) per mol albumin

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

General description

Albumin gene is mapped to human chromosome 4q13.3. It is synthesized as a preproalbumin with 609 amino acids in the liver. The mature albumin comprising 585 residues, is generated by the cleavage of signal peptide. Albumin is the major circulating protein in serum. It comprises three domains (I, II and III), each of which is subdivided into two subdomains A and B. A total of six ligand binding site is associated with albumin. Domain IIA and IIIA harbor small heterocyclic or aromatic compounds binding site. Long-chain fatty acid binding sites is present in IB and IIIB domains.

Application

Naturally occurring glycated albumin is a sensitive indicator of glycemic control by diabetic patients.
Albumin, glycated human has been used:
  • as a component of serum-free medium for retinal microglia activation
  • to stimulate immortalised proximal tubule epithelial (HK-2) cells for fibronectin production
  • as medium supplement in human retinal microvascular endothelial cells (HRMECs) cultures to test its effect on family with sequence similarity 18, member B (FAM18B) expression


Biochem/physiol Actions

Albumin is crucial for physiological processes and is the major transporter for lipids, metal ions and amino acids. It has antioxidant functionality and binds to drugs as well. Mutation in the albumin gene is implicated in an autosomal recessive disorder called Analbuminemia, which is characterized by low levels of albumin. The major glycation site is lysine 525 in albumin. Glycated albumin is a marker of glycemia in diabetes mellitus patients and is useful in assessment of HbA1c based glycemic control.

Physical form

Lyophilized powder containing citrate buffer salts

Preparation Note

Glycated in vitro

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A72231012595A6661
description

lyophilized powder

description

recombinant, expressed in Pichia pastoris, 5% in aqueous buffer, ≥90% (SDS-PAGE)

description

United States Pharmacopeia (USP) Reference Standard

description

lyophilized powder

assay

90-100% protein basis (biuret)

assay

≥90% (SDS-PAGE)

assay

-

assay

-

biological source

human

biological source

human

biological source

-

biological source

human

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

RNA extraction: suitable

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

200

form

lyophilized powder

form

solution

form

liquid

form

lyophilized powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

1–6 of 6 Questions  
  1. How many freeze/thaw cycles are permissible? Can aqueous solutions be mixed and then frozen for later use?

    1 answer
    1. Repeated freeze/thaw cycles are not recommended for this product; ideally, only one freeze-thaw cycle (for aliquoting) should be performed. Aqueous solutions can be mixed and then frozen for later use, but should be stored in single-use aliquots to avoid repeated freeze/thaw cycles. Stability in solution is not fully determined for this product, but similar products are stable for several months at –20°C.

      Helpful?

  2. How should this product be stored once reconstituted?

    1 answer
    1. The stability of this product in solution has not been determined. Non-glycated Human Serum Albumin (HSA) solution products are stable for at least 1 year when filter-sterilized and stored refrigerated.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What is the carbohydrate used for the glycation step of Product A8301, Albumin, glycated human?

    1 answer
    1. The starting carbohydrate for the albumin modification is glucose.

      Helpful?

  5. How much is Product A8301, Albumin, glycated human, glycated?

    1 answer
    1. The degree of glycation varies with each individual lot.  This value can be found on the Certificate of Analysis for each lot.

      Helpful?

  6. What is the molecular weight of Product A8301, Albumin, glycated human?

    1 answer
    1. Because the degree of glycation varies with each lot, the calculated molecular weight (MW) varies with each lot.  The parent albumin has a molecular weight of approximately 66,478 Daltons. Each hexose unit adds approximately 180 Daltons to the molecular weight.

      Helpful?

Reviews

No rating value

Active Filters