Skip to Content
Merck
All Photos(1)

Key Documents

A7986

Sigma-Aldrich

Atovaquone

≥98% (HPLC), powder, anti-protozoal agent

Synonym(s):

Mepron, trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19ClO3
CAS Number:
Molecular Weight:
366.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Atovaquone, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >10 mg/mL

originator

GlaxoSmithKline

storage temp.

−20°C

SMILES string

OC1=C([C@H]2CC[C@@H](CC2)c3ccc(Cl)cc3)C(=O)c4ccccc4C1=O

InChI

1S/C22H19ClO3/c23-16-11-9-14(10-12-16)13-5-7-15(8-6-13)19-20(24)17-3-1-2-4-18(17)21(25)22(19)26/h1-4,9-13,15,26H,5-8H2/t13-,15-

InChI key

KUCQYCKVKVOKAY-CTYIDZIISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Atovaquone inhibits the cytochrome bc(1) complex via interactions with the Rieske iron-sulfur protein and cytochrome b in the ubiquinol oxidation pocket. In addition to its use as a treatment for toxoplasmosis, atovaquone has antimalarial properties and prevents pneumocystis pneumonia post-renal transplant.

Biochem/physiol Actions

Atovaquone is an anti-protozoal mitochondrial electron transport inhibitor; Antimalarial; Antipneumocystic, and has also been used to treat toxoplasmosis. It is an analog of protozoan mitochondrial protein ubiquinone, and acts by inhibiting the cytochrome bc(1) complex via interactions with the Rieske iron-sulfur protein and cytochrome b in the ubiquinol oxidation pocket.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai J Rogers et al.
Cell reports, 30(12), 4041-4051 (2020-03-27)
During the 2013-2016 Ebola virus (EBOV) epidemic, a significant number of patients admitted to Ebola treatment units were co-infected with Plasmodium falciparum, a predominant agent of malaria. However, there is no consensus on how malaria impacts EBOV infection. The effect
Steven Gabardi et al.
Clinical transplantation, 26(3), E184-E190 (2012-04-11)
Pneumocystis pneumonia (PCP) is associated with significant morbidity and mortality in renal transplant recipients (RTR). Trimethoprim-sulfamethoxazole (TMP-SMZ) is considered the prophylactic agent-of-choice. Some patients require an alternative owing to TMP-SMZ intolerance. This is the first evaluation of full-dose atovaquone vs.
M Cella et al.
Clinical pharmacology and therapeutics, 91(4), 726-733 (2012-03-09)
Concurrent prescription of different drugs is common and is often necessary in many pediatric indications. A randomized concentration-controlled trial (RCCT) is proposed for pediatric studies in which drug combinations are used. The aim of our investigation was to show the
Zehua Song et al.
Antimicrobial agents and chemotherapy, 59(7), 4053-4058 (2015-04-29)
The bc1 complex is central to mitochondrial bioenergetics and the target of the antimalarial drug atovaquone that binds in the quinol oxidation (Qo) site of the complex. Structural analysis has shown that the Qo site residue Y279 (Y268 in Plasmodium
Mikio Kimura et al.
Parasitology international, 61(3), 466-469 (2012-04-10)
Malaria remains an important health risk among travelers to tropical/subtropical regions. However, in Japan, only 2 antimalarials are licensed for clinical use - oral quinine and mefloquine. The Research Group on Chemotherapy of Tropical Diseases introduced atovaquone-proguanil in 1999, and

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service