Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C4H10NO5P
CAS Number:
Molecular Weight:
183.10
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Form:
powder
form
powder
Quality Segment
optical purity
optical purity: ≥95% (HPLC, Marfey′s reagent)
SMILES string
N[C@@H](CCP(O)(O)=O)C(O)=O
InChI
1S/C4H10NO5P/c5-3(4(6)7)1-2-11(8,9)10/h3H,1-2,5H2,(H,6,7)(H2,8,9,10)/t3-/m0/s1
InChI key
DDOQBQRIEWHWBT-VKHMYHEASA-N
Gene Information
human ... GRM1(2911), GRM2(2912), GRM3(2913), GRM4(2914), GRM6(2916), GRM7(2917), GRM8(2918)
rat ... Grm1(24414), Grm2(24415), Grm4(24417), Grm5(24418), Grm6(24419), Grm7(81672)
Biochem/physiol Actions
mGluR4 and mGluR6 metabotropic glutamate receptor agonist.
Still not finding the right product?
Explore all of our products under
Storage Class
11 - Combustible Solids
wgk
WGK 3
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.