Skip to Content
Merck

A7736

Albumin human

recombinant, expressed in Pichia pastoris, lyophilized powder, ≥90% (agarose gel electrophoresis)

Synonym(s):

Albagen, rHSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C2936H4624N786O889S41
CAS Number:
Molecular Weight:
66471.53
UNSPSC Code:
12352202
NACRES:
NA.25
EC Number:
274-272-6
MDL number:
eCl@ss:
42010201
Biological source:
human
Assay:
≥90% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
cell culture | mammalian: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in Pichia pastoris

assay

≥90% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer ~67 kDa

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

General description

Construct with a single deletion of Asp from the N-terminus to create a hypoallergenic construct. Structural equivalence to native albumin confirmed x-ray crystallography. The Asp deletion eliminates an important antigenic determinant by eliminating the principal copper and nickel binding site of albumin. The first three residues of human albumin are considered non-structural. Also blocked at Cys34 to improve stability, monomer content and homogeneity.

Application

This recombinant variant is recommended for use in absorption, distribution, metabolism, and excretion (ADME) pharmacological research; cell culture; drug delivery research; and cryopreservation of cells.

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.

Other Notes

View more information on human serum albumin.

Legal Information

Albagen is a trademark of Albumin Therapeutics, LLC.


Still not finding the right product?

Explore all of our products under Albumin human


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library