Skip to Content
Merck

A7611

Azelastine hydrochloride

≥98% (HPLC)

Synonym(s):

4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)phthalazin-1-one hydrochloride, Astelin, Optivar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H24ClN3O · HCl
CAS Number:
Molecular Weight:
418.36
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

Wallace

storage temp.

−20°C

SMILES string

Cl.CN1CCCC(CC1)N2N=C(Cc3ccc(Cl)cc3)c4ccccc4C2=O

InChI

1S/C22H24ClN3O.ClH/c1-25-13-4-5-18(12-14-25)26-22(27)20-7-3-2-6-19(20)21(24-26)15-16-8-10-17(23)11-9-16;/h2-3,6-11,18H,4-5,12-15H2,1H3;1H

InChI key

YEJAJYAHJQIWNU-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Application

Azelastine hydrochloride is a H1 histamine receptor antagonist and NF-kB activator. Azelastine hydrochloride has been used in a study to investigate the potential of polymeric microspheres for treatment of allergic conjunctivitis.

Biochem/physiol Actions

H1 histamine receptor antagonist; NF-kB activator.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wallace. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Azelastine hydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library