Skip to Content
Merck
All Photos(3)

Key Documents

A7505

Sigma-Aldrich

1,4-Androstadiene-3,17-dione

Synonym(s):

1-Dehydroandrostenedione, Androstadienedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O2
CAS Number:
Molecular Weight:
284.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥85% (HPLC)

form

powder

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

chloroform: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])CC[C@]4(C)C(=O)CC[C@@]24[H]

InChI

1S/C19H24O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h7,9,11,14-16H,3-6,8,10H2,1-2H3/t14-,15-,16-,18-,19-/m0/s1

InChI key

LUJVUUWNAPIQQI-QAGGRKNESA-N

Gene Information

human ... CYP19A1(1588)

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Androstadiene-3,17-dione is a 17-keto[1] anabolic steroid.[2]

Biochem/physiol Actions

1,4-Androstadiene-3,17-dione is a prohormone that converts to an active steroid through the 17bHSD enzyme. 1,4-Androstadiene-3,17-dione is a metabolite of progesterone.
1,4-Androstadiene-3,17-dione is useful in forming pharmaceutically important steroids. Regio- and stereospecific hydroxylation of 1,4-androstadiene-3,17-dione increases its biological activity. Its derivative is also used to produce high-value bile acids.[3] 1,4-Androstadiene-3,17-dione serves as a precursor for the synthesis of progesterone, testosterone, cortisol, estradiol, cortisone, prednisolone, and prednisone.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa M Lopparelli et al.
Steroids, 76(5), 508-516 (2011-02-08)
Recently, the effect of illicit growth promoters (GPs) upon the cattle transcriptome has drawn the increasing attention of the scientific community. In the present study, the pre-transcriptional effects of three different illicit protocols on a set of target genes, including
Rosa Draisci et al.
Proteomics, 7(17), 3184-3193 (2007-08-07)
Surveillance of illegal use of steroids hormones in cattle breeding is a key issue to preserve human health. To this purpose, an integrated approach has been developed for the analysis of plasma and urine from calves treated orally with a
Francesca T Cannizzo et al.
The Journal of veterinary medical science, 69(11), 1109-1116 (2007-12-07)
Boldenone and its precursor Boldione are illegally used for anabolic purposes in humans, horses and cattle. To develop more effective policies and programs to maximize food security, Italian Public Health Services investigate all indicators capable of assisting the recognition of
M V Donova et al.
Microbiology (Reading, England), 153(Pt 6), 1981-1992 (2007-05-29)
Modified beta-cyclodextrins have been shown previously to enhance sterol conversion to 4-androstene-3,17-dione (AD) and 1,4-androstadiene-3,17-dione (ADD) by growing Mycobacterium spp. The enhancement effect was mainly attributed to steroid solubilization by the formation of inclusion complexes with modified cyclodextrins. In this
Wei Wei et al.
Applied and environmental microbiology, 76(13), 4578-4582 (2010-05-11)
3-Ketosteroid-Delta(1)-dehydrogenase, KsdD(M), was identified by targeted gene disruption and augmentation from Mycobacterium neoaurum NwIB-01, a newly isolated strain. The difficulty of separating 4-androstene-3,17-dione (AD) from 1,4-androstadiene-3,17-dione (ADD) is a key bottleneck to the microbial transformation of phytosterols in industry. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service