Skip to Content
Merck
All Photos(1)

Key Documents

A6185

Sigma-Aldrich

Calpain Inhibitor I

≥97% (TLC), powder

Synonym(s):

ALLN, Ac-LLnL-CHO, MG-101, N-Acetyl-L-leucyl-L-leucyl-L-norleucinal, N-Acetyl-Leu-Leu-Norleu-al

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37N3O4
CAS Number:
Molecular Weight:
383.53
Beilstein:
7656053
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥97% (TLC)

form

powder

color

white

solubility

DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

[H]C(=O)[C@H](CCCC)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(C)=O

InChI

1S/C20H37N3O4/c1-7-8-9-16(12-24)22-19(26)18(11-14(4)5)23-20(27)17(10-13(2)3)21-15(6)25/h12-14,16-18H,7-11H2,1-6H3,(H,21,25)(H,22,26)(H,23,27)/t16-,17-,18-/m0/s1

InChI key

FMYKJLXRRQTBOR-BZSNNMDCSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Calpain Inhibitor I have been used:
  • as a component of protease inhibitor cocktail for gel shift analysis
  • for treating HeLa cell cultures
  • for inhibition of 26 S proteasome in 26 S proteasome inhibition assays

Biochem/physiol Actions

Calpain Inhibitor I inhibits tumor necrosis factor-induced (TNF-induced) cleavage of Poly (ADP-ribose) polymerase (PARP) and calpastatin in apoptosis. It also inhibits oxygen/glucose deprivation (OGD) induced cleavage of caspase-12 and Bcl-xL..
Can initiate apoptosis in HL-60 cells but blocks dexamethasone-induced apoptosis in thymocytes and cycloheximide-induced apoptosis in metamyelocytes. Inhibits cyclin B degradation and arrests the cell cycle at G1/S and at meta-/anaphase. Blocks induction of nitric oxide synthase by LPS in macrophages.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Griscavage et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(8), 3308-3312 (1996-04-16)
The objective of this study was to elucidate the role of the proteasome pathway or multicatalytic proteinase complex in the induction of immunologic nitric oxide (NO) synthase (iNOS) in rat alveolar macrophages activated by lipopolysaccharide. Macrophages were incubated in the
Cleavage of the calpain inhibitor, calpastatin, during apoptosis
Porn-Ares M I, et al.
Cell Death and Differentiation, 5(12), 1028-1028 (1998)
Q Lu et al.
Archives of biochemistry and biophysics, 334(1), 175-181 (1996-10-01)
Recent investigations indicate that proteolysis is an important event in generation of the apoptosis phenotype. Although various proteases have been suggested to be candidates for this proteolysis, the results from different laboratories are inconsistent. In the present studies, HL-60 cells
S W Sherwood et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(8), 3353-3357 (1993-04-15)
The cytotoxic neutral cysteine protease inhibitor N-acetylleucylleucylnorleucinal (ALLN) inhibits cell-cycle progression in CHO cells, affecting the G1/S and metaphase-anaphase transition points, as well as S phase. Mitotic arrest induced by ALLN is associated with the inhibition of cyclin B degradation.
Cross-talk between two cysteine protease families: activation of caspase-12 by calpain in apoptosis
Nakagawa T and Yuan J
The Journal of Cell Biology, 150(4), 887-894 (2000)

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service