Skip to Content
Merck
All Photos(1)

Key Documents

A5168

Sigma-Aldrich

Adenosine Deaminase from bovine spleen

Type IX, ammonium sulfate suspension, 150-200 units/mg protein

Synonym(s):

Adenosine aminohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine spleen

Quality Level

type

Type IX

form

ammonium sulfate suspension

specific activity

150-200 units/mg protein

mol wt

32.5-33 kDa

UniProt accession no.

foreign activity

5′-AMP deaminase ≤0.01%
alkaline phosphatase ≤0.01%
guanase ≤0.01%
nucleoside phosphorylase ≤0.1%

storage temp.

2-8°C

Gene Information

cow ... ADA(280712)

Looking for similar products? Visit Product Comparison Guide

General description

Adenosine deaminase/ADA is an enzyme, that is mainly present in lymphocytes and monocytes.
Molecular weight: 32.5-33 kDa

pI: 4.85

Application

Adenosine Deaminase from bovine spleen has been used to metabolize adenosine triphosphate (ATP) and adenosine to an inactive form (inosine). It has also been used to as an additive in KRH (Krebs−Ringer solution buffered with Hepes, containing CaCl2) medium in order to remove exogenous adenosine for cyclic adenosine monophosphate (cAMP) assay.
Adenosine deaminase is useful in various molecular biology assays, such as glycerol release assays . Adenosine deaminase is a potential target for treatments of combined immunodeficiency disease.

Biochem/physiol Actions

Adenosine deaminase is a purine catabolic enzyme which catalyzes the deamination of adenosine and 2′-deoxyadenosine with approximately equal specificity. It is ubiquitous in mammalian tissue, and deficiency in adenosine deaminase has been associated with severe combined immunodeficiency disease.
Adenosine deaminase/ADA can be used as an important marker to identify tuberculous pleural effusion. The activity of ADA is usually high in liver disease, tuberculosis, typhoid, infective mononucleosis and several malignancies.

Unit Definition

One unit will deaminate 1.0 μmole of adenosine to inosine per min at pH 7.5 at 25 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4, 0.01 M potassium phosphate, pH 6.0

Analysis Note

Protein determined by biuret.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Paracrine stimulation of surfactant secretion by extracellular ATP in response to mechanical deformation
Patel A S, et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 289(3), L489-L496 (2005)
The diagnostic value of adenosine deaminase activity in pulmonary tuberculosis: Comparison between sputum and serum
Atta S, et al.
The Egyptian journal of chest diseases and tuberculosis, 64(1), 103-107 (2015)
Reduced PDE4 expression and activity contributes to enhanced catecholamine-induced cAMP accumulation in adipocytes from FOXC2 transgenic mice
Gronning L M, et al.
Febs Letters, 580(17), 4126-4130 (2006)
Ying Cheng et al.
Diabetes, 59(1), 17-25 (2009-10-17)
White adipose tissue (WAT) and brown adipose tissue (BAT) play distinct roles in adaptation to changes in nutrient availability, with WAT serving as an energy store and BAT regulating thermogenesis. We previously showed that mice maintained on a leucine-deficient diet
Mary Carmen Pérez-Aguilar et al.
Investigacion clinica, 53(3), 315-324 (2012-12-20)
Adenosine deaminase is an enzyme of the purine metabolism whose function is to convert adenosine to inosine and deoxyadenosine to deoxyinosine. The ecto-ADA1 binding to the cell surface through CD26 contributes to the regulation of cytokines and stimulates the proliferation

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service