Skip to Content
Merck

Skip To

A4700

Monoclonal Anti-Actin antibody produced in mouse

clone AC-40, ascites fluid

Synonym(s):

Monoclonal Anti-Actin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
AC-40, monoclonal
Application:
ELISA (i), IF, IHC (f), WB
Citations:
780
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

AC-40, monoclonal

mol wt

antigen 42 kDa

contains

15 mM sodium azide

species reactivity

human, Xenopus, hamster, mouse, guinea pig, rat, rabbit, snail, sheep, canine, carp, chicken, pig, bovine, viper, goat

technique(s)

immunohistochemistry (frozen sections): suitable using methanol-fixed, frozen sections, indirect ELISA: suitable, indirect immunofluorescence: 1:200 using cultured human or chicken fibroblasts, western blot: 1:500 using cultured human or chicken fibroblast extract

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Actin and myosin are major cytoskeletal proteins. Actin is expressed majorly in six isoforms. The human γ-actin gene is mapped to human chromosome 17.
The antibody recognizes an epitope located on the C-terminal end of actin conserved in all actin isoforms. It specifically labels actin in a wide variety of tissues and species. The epitope recognized by the antibody is resistant to formalin-fixation and paraffin-embedding. Zinc-formalin, B5, ethanol, methacarn, Brunnel′s or Bouin′s solutions may also be used as fixatives.

Monoclonal Anti-Actin (mouse IgG2a isotype) is derived from the AC-40 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. A synthetic actin C-terminal peptide attached to Multiple Antigen Peptide (MAP) backbone was used as the immunogen. The isotype is determined using Mouse Monoclonal Antibody Isotyping Reagents, Catalog No. ISO2.
The antibody recognizes an epitope located on the C-terminal end of actin conserved in all actin isoforms. It specifically labels actin in a wide variety of tissues and species. The epitope recognized by the antibody is resistant to formalin-fixation and paraffin-embedding. Zinc-formalin, B5, ethanol, methacarn, Brunnel′s or Bouin′s solutions may also be used as fixatives.
The two major cytoskeletal proteins implicated in cell motility are actin and myosin. Actin and myosin are constituents of many cell types and are involved in cellular process including locomotion, secretion, cytoplasmic streaming, phagocytosis, and cytokinesis. Although actin is one of the most conserved eukaryotic proteins, it is expressed in mammals and birds as at least six isoforms characterized by electrophoresis and amino acid sequence analysis.

Immunogen

synthetic actin C-terminal peptide, attached to a Multiple Antigen Peptide (MAP) backbone

Application

Monoclonal anti-actin antibody was used for western blot analysis of Cos-7 cell lysates to ensure equal protein loading.
Monoclonal rabbit anti-actin has been used:
  • in the detection of actin in HeLa cells, breast cancer cell line and leukemia cell line using western blotting
  • in human fibroblasts by immunofluorescence detection
Western blot analysis of MDCK cell lysates were performed using monoclonal anti-actin antibody as a primary antibody.

Biochem/physiol Actions

Actin and myosin are constituents of many cell types and are involved in cellular process including locomotion, secretion, cytoplasmic streaming, phagocytosis, and cytokinesis. The N-terminal region of actin appears to be a major antigenic region, and may be involved in the interaction of actin with other proteins such as myosin. Among the six isoforms of actin, four of them represent the differentiation markers of muscle tissues and two are found practically in all cells. There are three α-actins (skeletal, cardiac, and smooth muscle), one β−actin (β-nonmuscle), and two γ-actins (γ-smooth muscle and γ-nonmuscle). Actin isoforms show >90% overall sequence homology, but only 50–60% homology in their NH2-terminal residues.
The actin in cells of various species and tissues are very similar in their immunological and physical properties characterized by electrophoresis and amino acid sequence analysis.(from bulletin) They mediate cytoskeletal remodeling during T cell activation. Mutation in the γ-actin and β-actin gene is associated with a developmental disorder called Baraitser-Winter syndrome. Mutation in the γ-actin leads to hearing loss.
Monoclonal Anti-Actin recognizes an epitope located on the C-terminal end of actin, but not on the N-terminal end. This epitope is conserved in all actin isoforms. The antibody specifically labels actin in a wide variety of tissues and species in immunoblotting (band at 42 kDa), in immunofluorescent staining of cultured cell lines and in immunohistology. Cross-reaction has been observed with human, bovine, sheep, goat, pig, rabbit, dog, mouse, rat, guinea pig, hamster, chicken, carp, viper, frog, and snail. The antibody may be used in staining of methanol-fixed, frozen sections, and the epitope recognized by the antibody is resistant to formalin-fixation and paraffin-embedding. Zinc-formalin, B5, ethanol, methacarn, Brunnel′s or Bouin′s solution may also be used as fixatives.

Physical form

Supplied as ascites fluid with 15mM sodium azide as a preservative.

Preparation Note

For continuous use, store at 2-8 °C for a maximum of one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Other Notes

To view an Actin antibody selection guide, please visit www.sigmaaldrich.com/actin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2172F3046A2103
Gene Information

human ... ACTA1(58), ACTA1(58), ACTA1(58), ACTA2(59), ACTA2(59), ACTA2(59), ACTB(60), ACTC1(70), ACTC1(70), ACTC1(70), ACTG1(71), ACTG1(71), ACTG1(71), ACTG2(72), ACTG2(72), ACTG2(72)
mouse ... Acta1(11459), Acta1(11459), Acta1(11459), Acta2(11475), Acta2(11475), Acta2(11475), Actb(11461), Actb(11461), Actb(11461), Actc1(11464), Actc1(11464), Actc1(11464), Actg1(11465), Actg1(11465), Actg1(11465), Actg2(11468), Actg2(11468), Actg2(11468)
rat ... Acta1(29437), Acta1(29437), Acta1(29437), Acta2(81633), Acta2(81633), Acta2(81633), Actb(81822), Actb(81822), Actb(81822), Actc1(29275), Actc1(29275), Actc1(29275), Actg1(287876), Actg1(287876), Actg1(287876), Actg2(25365), Actg2(25365), Actg2(25365)

Gene Information

human ... ACTA1(58), ACTC1(70)
mouse ... Rwdd2a(69519)
rat ... Acta1(29437), Actc1(29275)

Gene Information

human ... ACTA1(58), ACTA1(58), ACTA1(58), ACTA2(59), ACTA2(59), ACTA2(59), ACTB(60), ACTC1(70), ACTC1(70), ACTC1(70), ACTG1(71), ACTG1(71), ACTG1(71), ACTG2(72), ACTG2(72), ACTG2(72)
mouse ... Acta1(11459), Acta1(11459), Acta1(11459), Acta2(11475), Acta2(11475), Acta2(11475), Actb(11461), Actb(11461), Actb(11461), Actc1(11464), Actc1(11464), Actc1(11464), Actg1(11465), Actg1(11465), Actg1(11465), Actg2(11468), Actg2(11468), Actg2(11468)
rat ... Acta1(29437), Acta1(29437), Acta1(29437), Acta2(81633), Acta2(81633), Acta2(81633), Actb(81822), Actb(81822), Actb(81822), Actc1(29275), Actc1(29275), Actc1(29275), Actg1(287876), Actg1(287876), Actg1(287876), Actg2(25365), Actg2(25365), Actg2(25365)

Gene Information

human ... ACTA1(58), ACTA2(59), ACTB(60), ACTC1(70), ACTG1(71), ACTG2(72)
mouse ... Acta1(11459), Acta2(11475), Actb(11461), Actc1(11464), Actg1(11465), Actg2(11468)
rat ... Acta1(29437), Acta2(81633), Actb(81822), Actc1(29275), Actg1(287876), Actg2(25365)

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit

clone

AC-40, monoclonal

clone

5C5, monoclonal

clone

AC-40, monoclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

FITC conjugate

conjugate

unconjugated

species reactivity

human, Xenopus, hamster, mouse, guinea pig, rat, rabbit, snail, sheep, canine, carp, chicken, pig, bovine, viper, goat

species reactivity

rabbit, carp, rat, sheep, guinea pig, human, snake, bovine, frog

species reactivity

human, Xenopus, hamster, mouse, guinea pig, rat, rabbit, snail, sheep, canine, carp, chicken, pig, bovine, viper, goat

species reactivity

frog, rat, mouse, chicken, human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters