Skip to Content
Merck

Skip To

A4679

Agarose

Low EEO, for Immunoelectrophoresis

Synonym(s):

3,6-Anhydro-α-L-galacto-β-D-galactan, Agarose LE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41105317
PubChem Substance ID:
EC Number:
232-731-8
NACRES:
NA.25
MDL number:
eCl@ss:
32110690
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (Gelidium)

Quality Segment

form

powder

technique(s)

electrophoresis: suitable, immunodiffusion: suitable, immunoelectrophoresis: suitable

impurities

≤7% water

EEO

0.09-0.13

mp

88 °C±2 °C (1.5% gel)

transition temp

gel point 34-38 °C (1.5% gel)

gel strength

≥1200 g/cm2 (1% gel)

anion traces

sulfate (SO42-): ≤0.20%

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

Agarose contains β-D-galactose and 3,6-anhydro-α-L-galactose, linked by glycosidic bonds β(1-4).

Application

Agarose is the most popular medium for immunoelectrophoresis because of the large pore size for rapid diffusion and for low background staining by Coomassie Blue G stain. The low EEO and high gel strength is specifically selected and tested for immunoelectrophoresis and immunodiffusion.

Preparation Note

Gel Preparation:
1. Prepare a 1% agarose solution (sufficient for 10 gels 85 mm x 100 mm, 1-1.5 mm thick) by mixing 1.5 g agarose (Catalog No. A4679) in 150 mL of prepared barbital buffer and heat in a boiling water bath until completely dissolved.
2. To prepare one gel, pour 14 mL of agarose solution onto the hydrophilic side of a level, well supported 85 mm x 100 mm sheet of Electrophoresis Film for Agarose Gels (Catalog No. E0264). Pour from the center of the sheet toward its edges forming an even layer of agarose 1-1.5 mm thick.
3. Allow the gels to harden for one hour at 4 °C before using or store at 0-5 °C in an appropriate, plastic wrapped container.

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A5030A0169A2790
description

Low EEO, for Immunoelectrophoresis

description

Ultra-low Gelling Temperature

description

Type I-A, low EEO

description

Wide range, Molecular Biology

form

powder

form

powder

form

powder

form

powder

gel strength

≥1200 g/cm2 (1% gel)

gel strength

-

gel strength

≥1200 g/cm2 (1% gel)

gel strength

-

EEO

0.09-0.13

EEO

≤0.05

EEO

0.09-0.13

EEO

≤0.15

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

technique(s)

immunodiffusion: suitable, immunoelectrophoresis: suitable

technique(s)

-

technique(s)

-

technique(s)

electrophoresis: suitable

impurities

≤7% water

impurities

-

impurities

≤7% water

impurities

-


Still not finding the right product?

Explore all of our products under Agarose


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters